Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0920-07

IMAC Sepharose High Performance

Cytiva 17-0920-07, pack of 100 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

packaging

pack of 100 mL

manufacturer/tradename

Cytiva 17-0920-07

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning in place

2-14(Medium uncharged with metal ions.)

working range

3-12

capacity

≥40 mg binding capacity (histidine-tagged protein/ml medium when changed with Ni2+)(Protein- and metal-ion dependent.)

General description

IMAC Sepharose High Performance media is easily charged by the user with the metalion of choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent purification of polyhistidine tagged proteins.

Application

Conveniently charge with your metal of choice. IMAC Sepharose FF is supplied free of metal ions. It is charged by the user with the transition metal ion of choice (e.g. Cu2?, Zn2?, Ni2?, or Co2?); these metal ions will bind to the covalently immobilized chelating ligand on the Sepharose. Small bead size for high performance purification and high binding capacity.

Features and Benefits

  • For optimizing purification of histidine-tagged Proteins when Ni2+ is not the best choice of metal ion
  • Conveniently charge with your metal of choice
  • 34 μm bead size for high performance purification
  • High binding capacity

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

IMAC Sepharose products, Ni Sepharose products and Fe Sepharose products
These products are sold under a license from Sigma-Aldrich under patent number EP 1276716 (Metal chelating compositions) and equivalent patents and patent applications in other countries.
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service