Skip to Content
Merck
All Photos(1)

Key Documents

GE11-0034-00

His Buffer Kit

Cytiva 11-0034-00

Synonym(s):

His Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
NA.25

sterility

non-sterile

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 11-0034-00

Related Categories

General description

His Buffer Kit contains carefully prepared buffer concentrates for binding, washing, and elution of histidine-tagged proteins according to recommended protocols. Moreover, method for optimization of imidazole concentration in the binding buffer is described.

Features and Benefits

His Buffer Kit can be used in combination with a number of column types, such as His SpinTrap, His MultiTrap FF, His MultiTrap HP, His GraviTrap, HisTrap FF, HisTrap FF crude and HisTrap HP

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HisTrap is a trademark of Cytiva
MultiTrap is a trademark of Cytiva

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service