Skip to Content
Merck
All Photos(1)

Key Documents

68268

Sigma-Aldrich

Imidazole buffer Solution

Imidazole buffer Solution

BioUltra, 1 M in H2O

Synonym(s):

Imidazole solution, Glyoxaline solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein:
103853
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

product line

BioUltra

Quality Level

form

liquid

concentration

1 M in H2O

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

9.5±0.5

density

1.01 g/mL at 20 °C

SMILES string

c1c[nH]cn1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

General description

Imidazole is used as a buffer in the pH range of 6.2-7.8 at 25°C. It is recommended as a component of a buffer for the assay of horseradish peroxidase and as a chelator for the binding of various divalent cations. Imidazole aids the elution of histidine-containing proteins from divalent cation resins (Sigma P6611, HisSelect.-HC Nickel affinity gel) and can also be used in reverse staining of SDS-PAGE gels for the detection of proteins.

Application

Imidazole buffer solution has been used as a component:
  • of calmodulin-binding buffer 1 (CBB1) for the tandem affinity purification (TAP) of TAP-tagged proteins together with co-purifying proteins
  • of lysis buffer, and SDS loading buffer for His-ubiquitin pull-down assays
  • of imidazole wash and elution buffer for protein purification

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Canning et al.
Journal of molecular biology, 426(13), 2457-2470 (2014-04-29)
The discoidin domain receptors (DDRs), DDR1 and DDR2, form a unique subfamily of receptor tyrosine kinases that are activated by the binding of triple-helical collagen. Excessive signaling by DDR1 and DDR2 has been linked to the progression of various human
Ali Tebbi et al.
The Journal of biological chemistry, 290(22), 14077-14090 (2015-04-17)
Ribonucleotide reductase (RnR) is a key enzyme synthesizing deoxyribonucleotides for DNA replication and repair. In mammals, the R1 catalytic subunit forms an active complex with either one of the two small subunits R2 and p53R2. Expression of R2 is S
Pam M Van Ry et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 23(8), 1285-1297 (2015-06-09)
Duchenne muscular dystrophy (DMD) is a fatal neuromuscular disease caused by mutations in the dystrophin gene, leading to the loss of a critical component of the sarcolemmal dystrophin glycoprotein complex. Galectin-1 is a small 14 kDa protein normally found in skeletal
Michael D Clark et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(28), E3669-E3678 (2015-07-01)
Acetylation is correlated with chromatin decondensation and transcriptional activation, but its regulation by histone deacetylase (HDAC)-bearing corepressor complexes is poorly understood. Here, we describe the mechanism of assembly of the mammalian Sin3L/Rpd3L complex facilitated by Sds3, a conserved subunit deemed
Ranjana Pal et al.
The Journal of biological chemistry, 290(37), 22520-22531 (2015-07-30)
CUX1 and CUX2 proteins are characterized by the presence of three highly similar regions called Cut repeats 1, 2, and 3. Although CUX1 is ubiquitously expressed, CUX2 plays an important role in the specification of neuronal cells and continues to

Questions

Reviews

Active Filters

    • Review 1
    • Votes 0
    5 out of 5 stars.

    Brilliant

    This is brilliant product.
    Save me a lot of time to prepare endotoxin free buffer.

    Helpful?

  1. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service