Key Documents
GBL724024
Grace Bio-Labs HybriSlip hybridization cover
L × W × thickness 24 mm × 40 mm × 0.25 mm, pkg of 1000 ea
Synonym(s):
coverslide, hydrophobic coverslip, nuclease free coverslip, thin plastic coverslip
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Pricing and availability is not currently available.
material
colorless
polycarbonate
packaging
pkg of 1000 ea
manufacturer/tradename
Grace Bio-Labs 724024
L × W × thickness
24 mm × 40 mm × 0.25 mm
General description
HybriSlip ideally suited for in situ hybridization, in situ PCR and hybridization to genomic arrays on glass slides.
HybriSlips are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. HybriSlips remain flat and will not curl, even at high temperatures.
  • RNase and DNase free
  • Hydrophobic
  • Disposable
  • Will not chip or break
Legal Information
HybriSlip is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)