Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs HybriSlip
™
hybridization cover
Products
Manufacturer
Length (mm)
Color
Sterility
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
GBL714022
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 22 mm × 40 mm × 0.25 mm, pkg of 100 ea
Expand
View Pricing
GBL714550
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 45 mm × 50 mm × 0.25 mm, pkg of 50 ea
Expand
View Pricing
GBL712222
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 22 mm × 22 mm × 0.25 mm, pkg of 100 ea
Expand
View Pricing
GBL716024
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 24 mm × 60 mm × 0.25 mm, pkg of 100 ea
Expand
View Pricing
GBL716022
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 22 mm × 60 mm × 0.25 mm, pkg of 100 ea
Expand
View Pricing
GBL726024
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 24 mm × 60 mm × 0.25 mm, pkg of 1000 ea
Expand
View Pricing
GBL726022
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 22 mm × 60 mm × 0.25 mm, pkg of 1000 ea
Expand
View Pricing
GBL724550
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 45 mm × 50 mm × 0.25 mm, pkg of 250 ea
Expand
View Pricing
GBL724024
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 24 mm × 40 mm × 0.25 mm, pkg of 1000 ea
Expand
View Pricing
GBL702430
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 24 mm × 30 mm × 0.25 mm, pkg of 100 ea
Expand
View Pricing
GBL714024
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 24 mm × 40 mm × 0.25 mm, pkg of 100 ea
Expand
View Pricing
GBL702525
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 25 mm × 25 mm × 0.25 mm, pkg of 1000 ea
Expand
View Pricing
GBL712525
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 25 mm × 25 mm × 0.25 mm, pkg of 100 ea
Expand
View Pricing
GBL722222
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 22 mm × 22 mm × 0.25 mm, pkg of 1000 ea
Expand
View Pricing
GBL724022
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 22 mm × 40 mm × 0.25 mm, pkg of 1000 ea
Expand
View Pricing
GBL722430
Grace Bio-Labs HybriSlip
™
hybridization cover,
L × W × thickness 24 mm × 30 mm × 0.25 mm, pkg of 1000 ea
Expand
View Pricing