G9637
Glycerol 3-phosphate Oxidase from Pediococcus sp.
lyophilized powder, 40-80 units/mg solid, pH 8.1
Synonym(s):
sn-Glycerol 3-phosphate:oxygen 2-oxidoreductase, L-Glycerol 3-phosphate Oxidase
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Recommended Products
biological source
bacterial (Pediococcus spp.)
Quality Level
form
lyophilized powder
specific activity
40-80 units/mg solid, pH 8.1
mol wt
~76 kDa (gel filtration)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Glycerol 3-phosphate Oxidase is a FAD-dependent α-glycerophosphate oxidase and catalyzes the formation of dihydroxyacetonephosphate. Glycerol 3-phosphate oxidase comprises of a flavin adenine dinucleotide (FAD) and a substrate binding domain and corresponds to molecular weight of 42 kDa. It exists as a monomer and the active site comprises of isoalloxazine ring and histidine 53, arginine 316 and lysine 345 residues crucial for ligand binding.
Application
Glycerol 3-phosphate Oxidase from Pediococcus sp. has been used in the kinetic assay to study gluconeogenesis in liver and in immobilization studies on pencil graphite (PG) electrode.
This enzyme is useful for enzymatic determination of triglyceride when coupled with lipoprotein lipase (LPL-311, LPL-314) and glycerokinase (GYK-301, GYK-311) in clinical analysis.
Physical properties
Isoelectric point : 4.1 -/+0.1
Michaelis constants : 3.2x10 -3M (L-α-Glycerophosphate), 6.8 x 10 -3M (D, L-form)
Inhibitors : Ionic detergents (SDS, LBS, etc.), Hg++, Ag+
Optimum pH : 8.0 - 8.5Optimum temperature : 35 - 40oC
pH Stability : pH 6.5 - 8.5 (25oC, 20hr) Thermal stability : below 40oC (pH 7.0, 15min)
Substrate specificity : The enzyme has the highest specificity for L-form of a-γlycerophosphate
Michaelis constants : 3.2x10 -3M (L-α-Glycerophosphate), 6.8 x 10 -3M (D, L-form)
Inhibitors : Ionic detergents (SDS, LBS, etc.), Hg++, Ag+
Optimum pH : 8.0 - 8.5Optimum temperature : 35 - 40oC
pH Stability : pH 6.5 - 8.5 (25oC, 20hr) Thermal stability : below 40oC (pH 7.0, 15min)
Substrate specificity : The enzyme has the highest specificity for L-form of a-γlycerophosphate
Unit Definition
One unit will oxidize 1.0 μmole of L-glycerol 3-phosphate to dihydroxyacetone phosphate with the formation of H2O2 per min at 37°C, at the appropriate pH.
Physical form
Lyophilized powder containing stabilizers
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Structure of alpha-glycerophosphate oxidase from Streptococcus sp.: a template for the mitochondrial alpha-glycerophosphate dehydrogenase
Colussi T, et al.
Biochemistry, 47(3), 965-977 (2008)
Characterization of glycerol phosphate oxidase from Streptococcus pneumoniae and its application for ketose synthesis
Li Z, et al.
Bioorganic & medicinal chemistry letters, 25(3), 504-507 (2015)
Effect of increasing levels of glycerin on growth rate, carcass traits and liver gluconeogenesis in young bulls
Ladeira MM, et al.
Anim. Feed Sci. Technol., 219, 241-248 (2016)
Fabrication of glycerol biosensor based on co-immobilization of enzyme nanoparticles onto pencil graphite electrode
Narwal V and Pundir CS
Analytical Biochemistry, 555, 94-103 (2018)
Crystal structure of the flavoenzyme PA4991 from Pseudomonas aeruginosa
Jacewicz A, et al.
Acta Crystallographica. Section F, Structural Biology Communications, 72(2), 105-111 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service