Skip to Content
Merck
All Photos(1)

Key Documents

G7548

Sigma-Aldrich

Penta-O-galloyl-β-D-glucose hydrate

≥96% (HPLC)

Synonym(s):

1,2,3,4,6-Penta-O-galloyl-b-D-glucopyranose, 1,2,3,4,6-pentakis(3,4,5-trihydroxybenzoate)-b-D-Glucopyranose, Gallic acid, pentaester with b-D-glucopyranose, Glucopyranose, pentagallate, b-D, PGG, Penta-1,2,3,4,6-O-galloyl-b-D-glucose, Penta-O-galloyl-beta-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H32O26 · xH2O
CAS Number:
Molecular Weight:
940.68 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥96% (HPLC)

form

powder

optical activity

[α]/D +14 to +26°, c = 0.2 in acetone-d6

color

light brown

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

O=C(O[C@H]([C@H]([C@@H]([C@@H](COC(C1=CC(O)=C(O)C(O)=C1)=O)O2)OC(C3=CC(O)=C(O)C(O)=C3)=O)OC(C4=CC(O)=C(O)C(O)=C4)=O)[C@@H]2OC(C5=CC(O)=C(O)C(O)=C5)=O)C6=CC(O)=C(O)C(O)=C6.O

InChI

1S/C41H32O26.H2O/c42-17-1-12(2-18(43)28(17)52)36(57)62-11-27-33(64-37(58)13-3-19(44)29(53)20(45)4-13)34(65-38(59)14-5-21(46)30(54)22(47)6-14)35(66-39(60)15-7-23(48)31(55)24(49)8-15)41(63-27)67-40(61)16-9-25(50)32(56)26(51)10-16;/h1-10,27,33-35,41-56H,11H2;1H2/t27-,33-,34+,35-,41+;/m1./s1

InChI key

JCJIMLXECAATFH-ZSJSDXHXSA-N

Application

Penta-O-galloyl-β-D-glucose hydrate (PCG) has been used as an inhibitor of abdominal aortic aneurysms (AAA) and matrix metalloproteinase (MMP) related metastatic activity.

Biochem/physiol Actions

PGG induces predominantly (caspase dependent) apoptosis in DU145 and LNCaP cells, while inducing autophagy in more resistant PC3 and TRAMP-C2 cells. It appears that PGG targeted signaling downstream of rather than the mTOR itself. Polyphenolic pyranoses were reported (J Biol Chem. 2010, 285 (11), 7892-902) to inhibit the plasminogen activator inhibitor type 1 (PAI-1).
Penta-O-galloyl-β-D-glucose hydrate (PCG) is a polyphenolic gallotannin compound produced by plants. It has an ability to inhibit matrix metalloproteinase (MMP) related metastatic activity. Thus, PCG can be used for treating metastatic activity in squamous cell carcinoma. In addition, it also acts as a potential drug for stabilizing small abdominal aneurysms.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Hui Lin et al.
Antimicrobial agents and chemotherapy, 55(3), 1021-1027 (2010-12-22)
1,2,3,4,6-Penta-O-galloyl-β-D-glucopyranose (PGG) is an active ingredient in plants that are commonly used in Chinese medicine to treat inflammation. We demonstrate here that PGG, at 6.25 μM, does not inhibit the growth of Staphylococcus aureus, and yet it prevents biofilm formation
Renate R Baumgartner et al.
Journal of natural products, 73(9), 1578-1581 (2010-09-02)
The inhibition of protein tyrosine phosphatase 1B (PTP1B) is of substantial interest for the treatment of type-2 diabetes mellitus. Using an in vitro enzyme assay with human recombinant PTP1B 1,2,3,4,6-penta-O-galloyl-D-glucopyranose was isolated from the roots of Paeonia lactiflora as an
Small molecule inhibition of matrix metalloproteinases as a potential therapeutic for metastatic activity in squamous cell carcinoma
Matias C, et al.
Oral cancer, 3(1-2), 1-8 (2019)
Hyo-Jung Lee et al.
Kidney international, 79(5), 538-545 (2010-11-19)
Adhesion of calcium oxalate (CaOx) crystals to kidney cells may be a key event in the pathogenesis of kidney stones associated with marked hyperoxaluria. Previously, we found that 1,2,3,4,6-penta-O-galloyl-β-D-glucose (PGG), isolated from a traditional medicinal herb, reduced CaOx crystal adhesion
Anna K Kiss et al.
Journal of agricultural and food chemistry, 58(18), 9960-9966 (2010-08-21)
In this study, we analyzed ex vivo the effect of an aqueous extract of Oenothera paradoxa defatted seeds on the formation of neutrophil-derived oxidants. For defining active compounds, we also tested lypophilic extract constituents such as gallic acid, (+)-catechin, ellagic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service