Skip to Content
Merck
All Photos(3)

Key Documents

C0692

Sigma-Aldrich

(−)-Catechin gallate

≥98% (HPLC), from green tea

Synonym(s):

(2S,3R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-(3,4,5-trihydroxybenzoate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18O10
CAS Number:
Molecular Weight:
442.37
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.77

biological source

green tea

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@@H](Oc2c1)c4ccc(O)c(O)c4

InChI

1S/C22H18O10/c23-11-6-14(25)12-8-19(32-22(30)10-4-16(27)20(29)17(28)5-10)21(31-18(12)7-11)9-1-2-13(24)15(26)3-9/h1-7,19,21,23-29H,8H2/t19-,21+/m1/s1

InChI key

LSHVYAFMTMFKBA-CTNGQTDRSA-N

Gene Information

human ... BACE1(23621)

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Catechin gallate belongs to a class of polyphenol compounds that are formed when catechins in green tea undergo epimerization on heat treatment. It is a naturally occurring flavanol catechin having a galloyl moiety.

Application

(−)-Catechin gallate has been used:
  • to examine the potency of catechins containing galloyl moiety in inhibiting the activity of human immunodeficiency virus-1 (HIV-1)integrase
  • to analyze the anti-oxidant and anti-viral properties of Pseudopiptadenia contorta and the commercial quebracho extracts in a herpes simplex virus type 1 strain, resistant to acyclovir
  • to study the effects of polyphenols on the biochemistry of human spermatozoa and the associated limitations of their use in gamete preservation

Biochem/physiol Actions

(−)-Catechin gallate inhibits the absorption of glucose in adipocytes isolated from rats. It is also reported to exhibit anti-oxidative properties along with a capacity to decrease plasma cholesterol levels. (−)-Catechin gallate shows the potency to be used as an agent to modify antibiotic resistance.
Antioxidant constituent of green tea.
Antioxidant constituent of green tea. At μM concentrations, it inhibits VEGF-induced tyrosine phosphorylation. It also inhibits aromatase activity, an enzyme that converts androgens to estrogen and is thought to play a role in the etiology of breast cancer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo Strobel et al.
The Biochemical journal, 386(Pt 3), 471-478 (2004-10-08)
The facilitative glucose transporter, GLUT4, mediates insulin-stimulated glucose uptake in adipocytes and muscles, and the participation of GLUT4 in the pathogenesis of various clinical conditions associated with obesity, visceral fat accumulation and insulin resistance has been proposed. Glucose uptake by
Meng Shi et al.
Food research international (Ottawa, Ont.), 132, 109050-109050 (2020-04-26)
Matcha-tuna oil and matcha-maltodextrin-tuna oil emulsions (25% oil, dry basis), formulated to have protein: carbohydrate ratios of 1:1.1, 1:2, 1:3 and 1:4, were spray dried. Confocal laser scanning microscopy showed effective emulsification of oil in all emulsions. All powders had
Antioxidant and antiviral properties of Pseudopiptadenia contorta (Leguminosae) and of quebracho (Schinopsis sp.) extracts
Moreira DL, et al.
Quimica nova, 28(3), 421-425 (2005)
Chuang Zhang et al.
Food chemistry, 244, 109-119 (2017-11-10)
Tea polyphenols have been a topic of discussion due to their health benefits. Nevertheless, detailed studies on the antioxidant capacity and polyphenol contents of teas in relation to factors including geographical locations, plantation elevations and leaf grades have been limited.
Shilpi Sharma et al.
Antimicrobial agents and chemotherapy, 51(7), 2552-2558 (2007-05-09)
The emergence of strains of Plasmodium falciparum resistant to the commonly used antimalarials warrants the development of new antimalarial agents. The discovery of type II fatty acid synthase (FAS) in Plasmodium distinct from the FAS in its human host (type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service