G6514
Gelatin veronal buffer
Synonym(s):
GVB2+, Gelatin Barbital Buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161700
NACRES:
NA.25
Recommended Products
form
liquid
Quality Level
drug control
Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IV (Portugal)
pH
7.3-7.4
storage temp.
2-8°C
Other Notes
0.15 mM CaCl2, 141 mM NaCl, 0.5 mM MgCl2, 0.1% gelatin, 1.8 mM sodium barbital and 3.1 mM barbituric acid, pH 7.3-7.4.
Aseptically filtered
Aseptically filtered
Preparation Note
Below 35-40 °C, gelatin swells in and absorbs 5-10 times its weight of water to form a gel. Gelatin is more soluble in hot than in cold water. If visible floaters or cloudiness is evident, solution may be warmed in 45°C water bath for 10 minutes to achieve clear, colorless solution.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Daniela M Ferreira et al.
Clinical and vaccine immunology : CVI, 15(3), 499-505 (2008-01-11)
We have previously shown that DNA immunization with PspA (pneumococcal surface protein A) DNA is able to elicit protection comparable to that elicited by immunization with PspA protein (with alum as adjuvant), even though the antibody levels elicited by DNA
Meghan A Morrissey et al.
Immunity, 53(2), 290-302 (2020-08-10)
CD47 acts as a "don't eat me" signal that protects cells from phagocytosis by binding and activating its receptor SIPRA on macrophages. CD47 suppresses multiple different pro-engulfment "eat me" signals, including immunoglobulin G (IgG), complement, and calreticulin, on distinct target
Nancy K Nyakoe et al.
Malaria journal, 8, 7-7 (2009-01-13)
Complement (C) can be activated during malaria, C components consumed and inflammatory mediators produced. This has potential to impair host innate defence. In a case-control study, C activation was assessed by measuring serum haemolytic activity (CH50), functional activity of each
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service