Skip to Content
Merck
All Photos(1)

Documents

B0300000

Barbital

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5,5-Diethyl-2,4,6(1H,3H,5H)-pyrimidinetrione, 5,5-Diethylbarbituric acid, Barbitone, Veronal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O3
CAS Number:
Molecular Weight:
184.19
Beilstein:
163999
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

barbital

manufacturer/tradename

EDQM

drug control

USDEA Schedule IV; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC1(CC)C(=O)NC(=O)NC1=O

InChI

1S/C8H12N2O3/c1-3-8(4-2)5(11)9-7(13)10-6(8)12/h3-4H2,1-2H3,(H2,9,10,11,12,13)

InChI key

FTOAOBMCPZCFFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Barbital EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Sedative/hypnotic

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Salcedo-Rivillas et al.
Clinical and vaccine immunology : CVI, 21(7), 972-981 (2014-05-09)
Pneumococcal surface protein A (PspA) is a candidate antigen for the composition of protein-based vaccines against Streptococcus pneumoniae. While searching for efficient adjuvants for PspA-based vaccines, our group has described the potential of combining PspA with the whole-cell pertussis vaccine
Kalana Maduwage et al.
PLoS neglected tropical diseases, 8(12), e3304-e3304 (2014-12-19)
Venom recurrence or persistence in the circulation after antivenom treatment has been documented many times in viper envenoming. However, it has not been associated with clinical recurrence for many snakes, including Russell's viper (Daboia spp.). We compare the recovery of
Megan Dodd et al.
The journal of gene medicine, 16(5-6), 131-142 (2014-06-21)
Hemophilia B patients are subject to frequent and spontaneous bleeding caused by a deficiency of clotting factor IX (FIX). Mesenchymal stromal cells (MSCs) have been used in cellular therapies as a result of their immunomodulatory properties, the ability to home
Juergen Patzke et al.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis, 25(8), 860-870 (2014-09-06)
The functional activity of von Willebrand factor (VWF) is most frequently measured by using the ristocetin cofactor assay (VWF:RCo). However, the method's drawbacks include unsatisfactory precision, sensitivity and availability of automated system applications. We have developed an alternative assay (INNOVANCE
Mahmoud Abdel-Latif et al.
International immunopharmacology, 24(2), 173-181 (2014-12-17)
Diethylcarbamazine citrate (DEC) had a significance in anti-filarial chemotherapy, while excretory-secretory product (ES) is released from adult filarial females. The target of the current study was to examine the immunomodulatory effect of DEC, Setaria equina ES or a combination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service