Skip to Content
Merck
All Photos(1)

Key Documents

G5921

Sigma-Aldrich

L-Glutamate Oxidase from Streptomyces sp.

recombinant, expressed in E. coli, lyophilized powder, ≥5.0 units/mg solid

Synonym(s):

L-Glutamate:oxygen oxidoreductase (deaminating)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

Quality Level

recombinant

expressed in E. coli

form

lyophilized powder

specific activity

≥5.0 units/mg solid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Glutamate is a non-essential amino acid.[1] It acts as a primary excitatory neurotransmitter.[2] In major trauma, major surgery, sepsis and bone marrow transplantation, glutamate functions as an essential amino acid.[1]

Application

L-Glutamate Oxidase from Streptomyces sp has been used to determine the enzyme activity of the immobilized glutamate oxidase (GOx).[3] It has also been used as a component of imaging buffer for stochastic optical reconstruction microscopy (STORM) imaging.[4]

Biochem/physiol Actions

L-glutamate oxidase catalyzes the conversion of L-glutamate to 2-oxoglutarate[5].
Major enzyme in the synthesis or degradation of glutamic acid; transfers an amine to α-ketoglutaric acid to form L-glutamic acid or deamidates L-glutamic acid

Unit Definition

One unit will form 1.0 μmole of α-ketoglutaric acid from L-glutamic acid per min at pH 7.4 at 30 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification and properties of a new enzyme, L-glutamate oxidase, from Streptomyces sp X-119-6 grown on wheat bran.
H. Yoshino
Agricultural and Biological Chemistry, 74, 1323-1328 (1983)
II. Glutamine and glutamate
Tapiero H, et al.
Biomedicine and Pharmacotherapy, 56(9), 446-457 (2002)
Dopamine and Glutamate in Psychiatric Disorders (2010)
Sensing based on the motion of enzyme-modified nanorods
Bunea AI, et al.
Biosensors And Bioelectronics, 67(9), 42-48 (2015)
Hong-Xuan Ren et al.
Talanta, 78(4-5), 1371-1377 (2009-04-14)
A novel matrix for enzyme immobilization was successfully developed by using Pt/Au bimetallic hierarchical structure with micro/nano-array to generate highly sensitive biosensors for glutamic oxaloacetic transaminase (GOT, EC 2.6.1.1) and glutamic pyruvic transaminase (GPT, EC 2.6.1.2) determination. The structures of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service