All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H21N3O5
CAS Number:
Molecular Weight:
275.30
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
γ-Glu-ε-Lys,
Assay
≥98.0% (TLC)
Quality Level
form
powder
color
white
storage temp.
−20°C
SMILES string
NC(CCCCNC(=O)CCC(N)C(O)=O)C(O)=O
InChI
1S/C11H21N3O5/c12-7(10(16)17)3-1-2-6-14-9(15)5-4-8(13)11(18)19/h7-8H,1-6,12-13H2,(H,14,15)(H,16,17)(H,18,19)
InChI key
JPKNLFVGUZRHOB-UHFFFAOYSA-N
Application
GammaGlu-epsilonLys (γ-Glu-ε-Lys) is used to study the functions and processing of endo-epsilon(-gamma-Glu)-Lys isopeptide bonds in biological processes.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chee-Yuen Gan et al.
International journal of food sciences and nutrition, 60 Suppl 7, 99-108 (2009-02-06)
Cross-linked soy protein isolate (SPI) gels were produced via single-treatment of SPI with microbial transglutaminase (MTG) for 5 h or 24 h, or with ribose for 2 h, or via combined-treatments of SPI with MTG followed by heating with ribose.
Tools for the detection and quantitation of protein transglutamination.
Zoltan Nemes et al.
Analytical biochemistry, 342(1), 1-10 (2005-06-17)
M A Rozenfel'd et al.
Izvestiia Akademii nauk. Seriia biologicheskaia, 6(6), 671-679 (2009-02-10)
The mechanism of enzymatic covalent crosslinking of fibrinogen molecules under the effect of plasma transglutaminase (factor XIIIa) has been studied. Using the methods of elastic and dynamic light scattering combined with viscosimetry and electrophoresis of the reduced samples, it has
Guylaine Hoffner et al.
Journal of mass spectrometry : JMS, 43(4), 456-469 (2007-12-08)
Protein aggregates are characteristic of a number of diseases of the central nervous system such as diseases of polyQ expansion. Covalent bonds formed by the action of transglutaminase are thought to participate in the stabilization of these aggregates. Transglutaminase catalyzes
Colin Robinson et al.
European journal of oral sciences, 119 Suppl 1, 50-54 (2012-01-25)
Tuft protein is a material associated with enamel tufts, and resides in dental enamel primarily at the enamel-dentine junction. It is located primarily at prism peripheries and extends in a very attenuated form towards the enamel outer surface. While it
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service