Skip to Content
Merck
All Photos(1)

Key Documents

G4166

Sigma-Aldrich

Glucose Isomerase from Streptomyces murinus

≥350 U/g

Synonym(s):

Sweetzyme® IT Extra, D-xylose ketol-isomerase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Streptomyces sp. (S. murinus)

Quality Level

form

powder

specific activity

≥350 U/g

storage temp.

2-8°C

General description

Immobilized glucose isomerase produced from a selected strain of Streptomyces murinus.

Application

Glucose Isomerase from Streptomyces murinus has been used to isomerize xylose to xylulose during the production of dihydrogen from Xylose. It has also been used in the synthetic enzymatic pathway for dihydrogen production from sucrose, to catalyze isomerization of glucose to fructose.
Immobilized glucose isomerase produced from Streptomyces murinus was used for the isomerization of xylose. Glucose isomerase is used in the food industry to produce high-fructose corn syrup.

Biochem/physiol Actions

Few anaerobic bacteria, fungi and plants express an intracellular metalloenzyme called D-xylose isomerase (XI). Most bacteria use the enzyme D-xylose isomerase to transform D-xylose to D-xylulose. D-Xylose isomerase (XI) converts the aldo-sugars xylose and glucose to their keto analogs xylulose and fructose.
Glucose isomerase has wide variety of industrial applications such as producing high-fructose corn syrup (HFCS) and production of ethanol from hemicelluloses. In addition, it also facilitates the study of structure-function relationships by advanced biochemical and genetic engineering techniques.

Physical properties

0.33 g yields an approximate bed volume of 1ml

Unit Definition

one unit converts glucose to fructose at an initial rate of 1 μmole per min at standard analytical conditions

Legal Information

A product of Novozyme Corp.
Sweetzyme is a registered trademark of Novozymes Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonardo de Figueiredo Vilela et al.
Bioresource technology, 128, 792-796 (2012-11-29)
This study presents results regarding the successful cloning of the bacterial xylose isomerase gene (xylA) of Burkholderia cenocepacia and its functional expression in Saccharomyces cerevisiae. The recombinant yeast showed to be competent to efficiently produce ethanol from both glucose and
Kripa Rao et al.
Applied biochemistry and biotechnology, 146(1-3), 101-117 (2008-04-19)
Of the sugars recovered from lignocellulose, D-glucose can be readily converted into ethanol by baker's or brewer's yeast (Saccharomyces cerevisiae). However, xylose that is obtained by the hydrolysis of the hemicellulosic portion is not fermentable by the same species of
Hiroki Kitayama et al.
Biochimica et biophysica acta, 1834(12), 2640-2646 (2013-10-08)
Protein crystals form in supersaturated solutions via a nucleation and growth mechanism. The amyloid fibrils of denatured proteins also form via a nucleation and growth mechanism. This similarity suggests that, although protein crystals and amyloid fibrils are distinct in their
Jürgen Behnen et al.
ChemMedChem, 7(2), 248-261 (2012-01-04)
Small highly soluble probe molecules such as aniline, urea, N-methylurea, 2-bromoacetate, 1,2-propanediol, nitrous oxide, benzamidine, and phenol were soaked into crystals of various proteins to map their binding pockets and to detect hot spots of binding with respect to hydrophobic
Linrui Wu et al.
Biomolecules, 10(7) (2020-07-16)
A fluorometabolite, 5-fluoro-5-deoxy-D-ribulose (5-FDRul), from the culture broth of the soil bacterium Streptomyces sp. MA37, was identified through a combination of genetic manipulation, chemo-enzymatic synthesis and NMR comparison. Although 5-FDRul has been chemically synthesized before, it was not an intermediate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service