Skip to Content
Merck

Skip To

FA20

Fructose Assay Kit

sufficient for 20 assays

Synonym(s):

Fructose Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12164500
EC Number:
200-333-3
NACRES:
NA.84
Technique(s):
photometry: suitable
Usage:
sufficient for 20 assays
Storage temp.:
2-8°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 20 assays

Quality Segment

analyte chemical class(es)

sugars (fructose)

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Application

This kit is for the quantitative, enzymatic determination of fructose in food and other materials.Fructose is phosphorylated by adenosine triphosphate (ATP) in the reaction catalyzed by hexokinase. Fructose 6-phosphate is converted to glucose 6-phosphate by phosphoglucose isomerase (PGI). Glucose-6-phosphate (G6P) is then oxidized to 6-phosphogluconate in the presence of nicotinamide adenine dinucleotide (NAD) in the reaction catalyzed by glucose-6-phosphate dehydrogenase (G6PDH). During this oxidation, an equimolar amount of NAD is reduced to NADH. The consequent increase in absorbance at 340 nm is directly proportional to fructose concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GAHK20TDF100ASA20
usage

sufficient for 20 assays

usage

sufficient for 20 assays

usage

sufficient for ~100 assays

usage

sufficient for 20 assays

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

analyte chemical class(es)

sugars (fructose)

analyte chemical class(es)

sugars (glucose)

analyte chemical class(es)

-

analyte chemical class(es)

sugars

application(s)

food and beverages
general analytical

application(s)

food and beverages
general analytical

application(s)

food and beverages
general analytical

application(s)

food and beverages
general analytical


Kit Components Only

Product No.
Description

  • fructose standard 500 mg

  • glucose assay reagent 1 mL/vial

  • phosphoglucose isomerase 1 mL/vial

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

wgk

WGK 3

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
FA20-1KT04061833622292
G3293-20ML04061837904981
G3293-50ML04061837904998
F2793-500MG04061833613313

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. イネの根および葉のフルクトース含量を測定したいと思っています。こちらのキットを使えるでしょうか。また、商品の詳細(分析手順等)について教えていただけると幸いです。

    1 answer
    1. Yes, this kit would be suitable. Here is the link to the Technical Bulletin that describes the use of this product:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/111/939/fa20bul.pdf
      For roots and leaves, one would follow the Sample Preparation protocol for Solids. The plant tissue would need to be macerated in order to completely extract the Fructose into the water.

      Helpful?

Reviews

No rating value

Active Filters