Skip to Content
Merck
All Photos(1)

Key Documents

G3295

Sigma-Aldrich

GW0742

≥98% (HPLC)

Synonym(s):

4-[2-(3-Fluoro-4-trifluoromethyl-phenyl)-4-methyl-thiazol-5-ylmethylsulfanyl]- 2-methyl-phenoxy}-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17F4NO3S2
CAS Number:
Molecular Weight:
471.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

mp

134.5-135.5 °C

solubility

DMSO: >5 mg/mL
H2O: insoluble

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

Cc1cc(SCc2sc(nc2C)-c3ccc(c(F)c3)C(F)(F)F)ccc1OCC(O)=O

InChI

1S/C21H17F4NO3S2/c1-11-7-14(4-6-17(11)29-9-19(27)28)30-10-18-12(2)26-20(31-18)13-3-5-15(16(22)8-13)21(23,24)25/h3-8H,9-10H2,1-2H3,(H,27,28)

InChI key

HWVNEWGKWRGSRK-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

GW0742 is a highly selective PPARδ agonist. EC50 = 1 nM vs 1 μM and 2 μM for PPARα and PPARγ, respectively.

Features and Benefits

This compound is featured on the Nuclear Receptors (PPARs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruce E Heck et al.
Cell transplantation, 26(8), 1405-1417 (2017-09-14)
Osteoarthritis (OA) is an inflammatory joint disease characterized by degeneration of articular cartilage within synovial joints. An estimated 27 million Americans suffer from OA, and the population is expected to reach 67 million in the United States by 2030. Thus
Amar Bahadur Singh et al.
The Journal of biological chemistry, 292(3), 884-897 (2016-12-04)
Peroxisome proliferator-activated receptor δ (PPARδ) regulates many genes involved in lipid metabolism. Hepatic lysophosphatidylcholine acyltransferase 3 (LPCAT3) has critical functions in triglycerides transport and endoplasmic reticulum stress response due to its unique ability to catalyze the incorporation of polyunsaturated fatty
Luka Opitz et al.
Journal of anatomy, 231(6), 970-977 (2017-08-09)
Pulmonary alveolar septa are thought to contain at least two types of fibroblasts that are termed myofibroblasts and lipofibroblasts based on their morphological characteristics. Lipofibroblasts possess cytoplasmic lipid inclusions (lipid bodies or droplets) and are involved in several important functions
Haruyo Yagi et al.
Stem cell research & therapy, 11(1), 293-293 (2020-07-19)
There have been limited advances in the treatment of bone and joint infections, which currently involves a combination of surgery and antibiotic administration. There is a timely need in orthopedics to develop more effective and less invasive forms of antimicrobial
Daniel Morales-Cano et al.
Clinical science (London, England : 1979), 130(20), 1823-1836 (2016-07-15)
PPARβ/δ activation protects against endothelial dysfunction in diabetic models. Elevated glucose is known to impair cAMP-induced relaxation and Kv channel function in coronary arteries (CA). Herein, we aimed to analyse the possible protective effects of the PPARβ/δ agonist GW0742 on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service