Skip to Content
Merck
All Photos(1)

Key Documents

G2295

Sigma-Aldrich

Glimepiride

≥98% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34N4O5S
CAS Number:
Molecular Weight:
490.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

mp

212.2-214.5 °C

solubility

DMSO: >10 mg/mL

originator

Sanofi Aventis

storage temp.

room temp

SMILES string

CCC1=C(C)CN(C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)N[C@H]3CC[C@H](C)CC3)C1=O

InChI

1S/C24H34N4O5S/c1-4-21-17(3)15-28(22(21)29)24(31)25-14-13-18-7-11-20(12-8-18)34(32,33)27-23(30)26-19-9-5-16(2)6-10-19/h7-8,11-12,16,19H,4-6,9-10,13-15H2,1-3H3,(H,25,31)(H2,26,27,30)/t16-,19-

InChI key

WIGIZIANZCJQQY-RUCARUNLSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Glimepiride is a class II biopharmaceutical second-generation sulfonylurea.

Application

Glimepiride has been used:
  • as a hypoglycemic drug to test its anti-diabetic functionality in human umbilical vein cells (HUVECs)
  • as a sulfonylurea ATP-sensitive potassium channel (KATP) channel inhibitor in breast cancer MDA-MB-231 cells
  • in glucose-stimulated insulin secretion (GSIS) assays of neonatal islet-like cell clusters (NICCs) to test its effect on insulin secretion

Glimepiride is currently used to treat type 2 diabetes.

Biochem/physiol Actions

Glimepiride is a potent blocker of cardiac KATP channels activated by pinacidil with an IC50 of 6.8 nM.
Glimepiride reduces blood glucose levels by stimulating the pancreatic β cells to secrete insulin hormone. It interacts with a 65-kD protein associated with β cells.

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Ping Wang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 38(6), 2337-2347 (2016-05-21)
By inducing severe endothelial impairment, hypertension and diabetes are two leading causes of morbidity and mortality. Hypertensive patients with concomitant diabetes must take both antihypertensive and hypoglycaemic medications, for which there is a lack of experimental and clinical guidelines. This
A REVIEW ARTICLE ON GLIMEPERIDE: AN ORAL HYPOGLYCAEMIC DRUG
Tiwari A, et al.
International Journal of Advanced Research , 4, 920-927 (2016)
Lan Gao et al.
International journal of technology assessment in health care, 28(4), 436-444 (2012-09-26)
The aim of this study was to evaluate the long-term cost-utility of liraglutide versus glimepiride as add-on therapy to metformin in patients with type 2 diabetes mellitus (T2DM), based on the results of clinical trial conducted in Asian population. The
INS-eGFP transgenic pigs: a novel reporter system for studying maturation, growth and vascularisation of neonatal islet-like cell clusters.
Elisabeth Kemter et al.
Diabetologia, 60(6), 1152-1156 (2017-03-21)
W Rathmann et al.
Diabetes, obesity & metabolism, 15(1), 55-61 (2012-08-07)
To investigate therapy persistence, frequency of hypoglycaemia and macrovascular outcomes among type 2 diabetes patients with dipeptidyl peptidase-4 (DPP-4) inhibitors (DPP-4) and sulphonylureas (SU). Data from 19,184 DPP-4 (mean age: 64 years; 56% males) and 31,110 SU users (69 years;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service