Skip to Content
Merck
All Photos(1)

Key Documents

Y0000516

Glimepiride for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Glimepiride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34N4O5S
CAS Number:
Molecular Weight:
490.62
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

glimepiride

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC1=C(C)CN(C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)N[C@H]3CC[C@H](C)CC3)C1=O

InChI

1S/C24H34N4O5S/c1-4-21-17(3)15-28(22(21)29)24(31)25-14-13-18-7-11-20(12-8-18)34(32,33)27-23(30)26-19-9-5-16(2)6-10-19/h7-8,11-12,16,19H,4-6,9-10,13-15H2,1-3H3,(H,25,31)(H2,26,27,30)/t16-,19-

InChI key

WIGIZIANZCJQQY-RUCARUNLSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glimepiride is currently used to treat type 2 diabetes.

Biochem/physiol Actions

Glimepiride is a potent blocker of cardiac KATP channels activated by pinacidil with an IC50 of 6.8 nM.
Potent blocker of cardiac KATP channels.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Mateos et al.
Drugs of today (Barcelona, Spain : 1998), 48 Suppl B, 1-17 (2012-10-20)
New drugs for type 2 diabetes that act on incretin metabolism have been shown to improve glycemic control, reduce body weight and have a low risk for hypoglycemia. Among these, liraglutide is the first glucagon-like peptide 1 (GLP-1) analogue approved
Shinzo Hiroi et al.
Diabetes technology & therapeutics, 15(2), 158-165 (2013-01-08)
This study aimed to determine the efficacy and safety of pioglitazone/glimepiride as a fixed-dose combination (FDC) in Japanese patients with type 2 diabetes. In this multicenter, phase III, open-label evaluation, eligible patients had to have a glycosylated hemoglobin (HbA(1c)) level
DPP-4 inhibition and neuroprotection: do mechanisms matter?
Richard P Shannon
Diabetes, 62(4), 1029-1031 (2013-03-23)
Mitsuyoshi Takahara et al.
Endocrine journal, 59(12), 1131-1136 (2012-08-02)
We retrospectively investigated the effect of adding dipeptidyl peptidase-4 (DPP-4) inhibitor and tapering sulfonylurea on blood glucose fluctuation in Asian patients with type 2 diabetes mellitus under basal-supported oral therapy (BOT). We recruited twenty-two consecutive Japanese patients with type 2
Elham A Mohamed et al.
AAPS PharmSciTech, 13(3), 1013-1023 (2012-07-31)
Effects of tromethamine (Tris), polyvinylpyrrolidone (PVP-K25), and low molecular weight chitosan (LM-CH) on dissolution and therapeutic efficacy of glimepiride (Gmp) were investigated using physical mixtures (PMs), coground mixtures, coprecipitates (Coppts) or kneaded mixtures (KMs), and compared with drug alone. Fourier

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service