Skip to Content
Merck
All Photos(1)

Key Documents

G1514

Sigma-Aldrich

D-(+)-Glucosamine hydrochloride

≥99%, BioReagent, suitable for cell culture

Synonym(s):

2-Amino-2-deoxy-D-glucose hydrochloride, Chitosamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein:
4157370
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.75

biological source

Aspergillus niger
maize fruit

Quality Level

product line

BioReagent

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

190-194 °C (dec.) (lit.)

solubility

water: 100 mg/mL, clear, colorless

SMILES string

Cl.N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3-,4-,5-,6?;/m1./s1

InChI key

QKPLRMLTKYXDST-NSEZLWDYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glucosamine, an amino sugar is generated form glucose and glutamine and is naturally produced in the body. Glucosamine, a endogenous amino monosaccharide is the major source of glucosamine-6-phosphate and n-acetylglucosamine.

Application

Glucosamine has been used as a standard to examine the concentration of glucosamine in the synovial fluid and its pharmacokinetics in serum.

Biochem/physiol Actions

Glucosamine is preferred as a nutritional supplement for individuals with osteoarthritis. It is used as a building block for the production of proteoglycans and glycosaminoglycans.
Glucosamine, an amino sugar, is the precursor of the hexosamine biosynthetic pathway leading to the formation of UDP-N-acetylglucosamine (UDP-GlcNAc), which is then used for making glycosaminoglycans, proteoglycans, and glycolipids. D-(+)-Glucosamine inhibits the coaggregation of Candida yeast species with the bacterial strain S. salivarius.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucosamine Chondroitin Sulfate
Saunders Handbook of Veterinary Drugs, 357-358 (2016)
Synovial fluid levels and serum pharmacokinetics in a large animal model following treatment with oral glucosamine at clinically relevant doses
Laverty S, et al.
Arthritis, 52(1), 181-191 (2005)
Nutrients and Dietary Supplements for Osteoarthritis
Bioactive Food as Dietary Interventions for Arthritis and Related Inflammatory Diseases, 97-137 (2019)
Sports Medicine and Concussion Management
Orthopaedic Physical Therapy Secrets, 172-182 (2017)
Nathaniel S Hwang et al.
Biomaterials, 27(36), 6015-6023 (2006-07-29)
Differentiation of embryonic stem (ES) cells generally occurs after formation of three-dimensional cell aggregates, known as embryoid bodies (EBs). We have previously reported that hydrogels provide EBs a supportive environment for in vitro chondrogenic differentiation and three dimensional tissue formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service