Skip to Content
Merck
All Photos(4)

Key Documents

G0668

Sigma-Aldrich

GW2974

≥98% (HPLC), solid

Synonym(s):

N4-(1-Benzyl-1H-indazol-5-yl)-N6,N6-dimethyl-pyrido­[3,4-d]­pyrimidine-4,6-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21N7
CAS Number:
Molecular Weight:
395.46
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

yellow

solubility

DMSO: ≥20 mg/mL

originator

GlaxoSmithKline

SMILES string

CN(C)c1cc2c(Nc3ccc4n(Cc5ccccc5)ncc4c3)ncnc2cn1

InChI

1S/C23H21N7/c1-29(2)22-11-19-20(13-24-22)25-15-26-23(19)28-18-8-9-21-17(10-18)12-27-30(21)14-16-6-4-3-5-7-16/h3-13,15H,14H2,1-2H3,(H,25,26,28)

InChI key

DYYZXRCFCVDSKD-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Potent and selective dual inhibitor of EGFR (IC50 = 0.016 mM) and ErbB-2 receptor tyrosine kinase.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Agarwal et al.
British journal of cancer, 100(6), 941-949 (2009-02-26)
There is a pressing need to identify new drug targets and novel approaches for treatment of non-small-cell lung carcinoma (NSCLC). Members of the epidermal growth factor receptor (EGFR) and Met receptor families have been identified as important molecular targets for
Keli Kolegraff et al.
Molecular biology of the cell, 22(8), 1121-1134 (2011-02-18)
Desmocollin-2 (Dsc2) and desmoglein-2 (Dsg2) are transmembrane cell adhesion proteins of desmosomes. Reduced expression of Dsc2 has been reported in colorectal carcinomas, suggesting that Dsc2 may play a role in the development and/or progression of colorectal cancer. However, no studies
Hauke Busch et al.
Molecular systems biology, 4, 199-199 (2008-07-03)
Translation of large-scale data into a coherent model that allows one to simulate, predict and control cellular behavior is far from being resolved. Assuming that long-term cellular behavior is reflected in the gene expression kinetics, we infer a dynamic gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service