FG0100
Free Glycerol Determination Kit
1 kit sufficient for 1000 reactions
Synonym(s):
Triglyceride and Free Glycerol Kits and Reagents
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
kit sufficient for 1000 reactions
detection method
colorimetric
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Glycerol, also referred to as glycerin or glycerine, is a 3 carbon sugar alcohol that forms the backbone of fatty acids. It is a central component for synthesis of all lipids, and acts as a backbone for triglycerides and phospholipids, which play an important role in cell membrane structure. Due to its low toxicity, glycerol is widely used in pharmaceutical, food and cosmetic formulations and is the main waste by-product of biodiesel production via transesterification. In addition to kits, the individual reagents and glycerol standard are available separately when fewer reactions are needed.
Application
The Free Glycerol Determination Kit has been used to determine the level of glycerol released into the medium from mammalian cells grown in vitro .
The Free Glycerol Determination Kit measures free, endogenous glycerol using coupled enzyme reactions and does not include initial lipase hydrolysis. Glycerol is phosphorylated by adenosine-5′-triphosphate (ATP) forming glycerol-1-phosphate (G-1-P) and adenosine-5′-diphosphate (ADP) in the reaction catalyzed by glycerol kinase (GK). G-1-P is then oxidized by glycerol phosphate oxidase (GPO) to dihydroxyacetone phosphate (DAP) and hydrogen peroxide (H2O2). Peroxidase catalyzes the coupling of H2O2 with 4-aminoantipyrine (4-AAP) and sodium N-ethyl-N-(3-sulfopropyl) m-anisidine (ESPA) to produce a quinoneimine dye that shows an absorbance maximum at 540 nm. The increase in absorbance at 540 nm is directly proportional to the free glycerol concentration of the sample.
The triglyceride and free glycerol kits are for the quantitative determination of glycerol, total triglycerides or free triglycerides.
Suitability
Suitable for the quantitative enzymatic determination of glycerol in serum or plasma.
Linkage
In addition to kits, the individual reagents and glycerol standard are available separately when fewer reactions are needed.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Obesity (Silver Spring, Md.), 25(10), 1707-1715 (2017-08-08)
To compare the individual and combined effects of dairy and endurance exercise training in reducing weight gain and adiposity in a rodent model of diet-induced obesity. An 8-week feeding intervention of a high-fat, high-sugar diet was used to induce obesity
EBioMedicine, 30, 248-260 (2018-03-28)
Long non-coding RNAs (lncRNAs) belong to a recently discovered class of molecules proposed to regulate various cellular processes. Here, we systematically analyzed their expression in human subcutaneous white adipose tissue (WAT) and found that a limited set was differentially expressed
Journal of clinical medicine, 9(7) (2020-07-28)
The relationship between liver disease and neuropathology in hepatic encephalopathy is well known, but the genesis of encephalopathy in liver failure is yet to be elucidated. Conceptually, the main cause of hepatic encephalopathy is the accumulation of brain ammonia due
Nutrition (Burbank, Los Angeles County, Calif.), 55-56, 63-70 (2018-07-01)
Non-alcoholic fatty liver disease is characterized by high hepatic triacylglycerol contents, which is associated with endoplasmic reticulum (ER) stress and insulin resistance. Caffeic acid (CA) has antioxidant, immunomodulatory, and antiinflammatory effects. We investigated the effects of CA on hepatic steatosis
Journal of lipid research, 51(12), 3455-3462 (2010-09-03)
Cidea, the cell death-inducing DNA fragmentation factor-α-like effector (CIDE) domain-containing protein, is targeted to lipid droplets in mouse adipocytes, where it inhibits triglyceride hydrolysis and promotes lipid storage. In mice, Cidea may prevent lipolysis by binding and shielding lipid droplets
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service