Skip to Content
Merck
All Photos(2)

Key Documents

F9911

Sigma-Aldrich

Fibronectin Proteolytic Fragment from human plasma

lyophilized powder, 30 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
42020141
NACRES:
NA.75

biological source

human plasma

Quality Level

Assay

>90% (SDS-PAGE)

form

lyophilized powder

mol wt

30 kDa

packaging

pkg of 0.5 mg

technique(s)

cell culture | mammalian: suitable

impurities

HIV and HBsAg, source material tested negative
Small proteolytic fragments, may contain traces

solubility

water: soluble >0.5 mg/mL, clear to slightly hazy, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... FN1(2335)

General description

Fibronectin 1 is a glycoprotein of the extracellular matrix that is coded by FN1 gene. It is expressed in the plasma and at the cell surface. FN1 is mapped to human chromosome 2q35. Proteolytic fragments of fibronectin has a cell binding domain.

Application

Fibronectin proteolytic fragment has been used in solid phase binding assay.

Biochem/physiol Actions

Fibronectin participates in cell adhesion, growth, migration, wound healing, blood coagulation and metastasis. Mutations in FN1 results in glomerulopathy. It plays an important role in cell attachment and spreading, control of cell cytoskeleton, morphology and differentiation. FN1 is also involved in extracellular matrix formation, hemostasis and thrombosis. Proteolytic fragments of fibronectin plays a vital role in mononuclear phagocyte function.

Caution

The product should be stored at -20°C.

Preparation Note

This product is lyophilized from phosphate buffered saline with sucrose as a cryoprotectant. The source material has tested negative for antibody to HIV, HCN, and HBsAg. It is soluble in water at 0.5 mg/mL and yields a clear to slightly hazy solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maryada Sharma et al.
PloS one, 8(7), e69343-e69343 (2013-08-13)
Fibrosis is characterized by excessive accumulation of scar tissue as a result of exaggerated deposition of extracellular matrix (ECM), leading to tissue contraction and impaired function of the organ. Fibronectin (Fn) is an essential component of the ECM, and plays
S Huff et al.
The Journal of biological chemistry, 269(22), 15563-15570 (1994-06-03)
The N-terminal 29-kDa fragment of fibronectin (Fn29K) contains five type I "finger" modules. It binds to heparin, fibrin, and bacteria and is involved in fibronectin (Fn) matrix assembly. Binding to Staphylococcus aureus involves a cell wall-associated protein that contains approximately
Fibronectin in Health and Disease (2018)
E Nègre et al.
The Journal of biological chemistry, 269(35), 22039-22045 (1994-09-02)
A 30-kDa proteolytic fragment from the gelatin/collagen-binding domain of fibronectin is a potent inhibitor of fibronectin binding to Candida albicans, with a molar inhibition constant equal to that of intact fibronectin. Recombinant and proteolytic fragments from the cell-, the fibrin
Ming Liu et al.
Scientific reports, 5, 14449-14449 (2015-09-25)
Bacterial adhesins play a pivotal role in the tight bacteria-host cells attachment to initiate the downstream processes and bacterial infection of hosts. In this study, we identified a novel adhesin, VpadF in V. parahaemolyticus. Deletion of VpadF in V. parahaemolyticus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service