Skip to Content
Merck
All Photos(1)

Key Documents

10838039001

Roche

Fibronectin

from human plasma

Synonym(s):

Fibronectin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207

biological source

human plasma

Quality Level

sterility

non-sterile; 0.2 μm filtered

Assay

95% (SDS-PAGE)

form

lyophilized (clear, colorless solution after reconstitution)

mol wt

440 kDa

packaging

pkg of 5 mg

manufacturer/tradename

Roche

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... FN1(2335)

Looking for similar products? Visit Product Comparison Guide

General description

Fibronectin is responsible for the cellular interactions with the extracellular matrix. It is involved in cell adhesion, migration, growth and differentiation. It is a soluble component of plasma and other body fluids. In addition, it is also part of the insoluble extracellular matrix.

Specificity

Active on most mammalian cells

Application

Fibronectin promotes the attachment and subsequent spreading of many cells. It is used for the coating of culture dishes.

Testing Method

Tested for the promotion of adherence of 3T3 (NR6) cells (XTT cleavage)

Physical form

Lyophilizate (contains 1.126 mg glycine and 0.058 mg sodium chloride per mg fibronectin), before lyophilization filtered through a 0.2 μm pore size membrane.

Preparation Note

Working concentration: 5 μg/cm2
5 μg/cm2 for the coating of cell culture vessels
Storage conditions (working solution): -15 to -25 °C
The reconstituted solution is stable at -15 to -25 °C.
Note: Store the reconstituted solution in aliquots at -15 to -25 °C.
Avoid repeated freezing and thawing.

Reconstitution

Add 1 or 5 ml sterile water, respectively, to yield a final concentration of 1 mg/ml. Incubate for 30 to 60 minutes at 37 °C to dissolve. Do not agitate.
Upon reconstitution, the solution may contain a small amount of insoluble aggregated material. These aggregates are a phenomenon inherent to fibronectin and do not influence product performance.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniella Penko et al.
Cell transplantation, 24(1), 37-48 (2013-09-28)
The success of pancreatic islet transplantation is limited by delayed engraftment and suboptimal function in the longer term. Endothelial progenitor cells (EPCs) represent a potential cellular therapy that may improve the engraftment of transplanted pancreatic islets. In addition, EPCs may
Hao Wang et al.
Microsystems & nanoengineering, 8, 49-49 (2022-05-14)
Cardiovascular disease is the number one cause of death in humans. Therefore, cardiotoxicity is one of the most important adverse effects assessed by arrhythmia recognition in drug development. Recently, cell-based techniques developed for arrhythmia recognition primarily employ linear methods such
Weiwei Wang et al.
Journal of oncology, 2022, 4499876-4499876 (2022-07-09)
Application of extracellular vesicles (EVs) for cancer treatment has been well-documented. We probed into the potential role of cervical cancer cells-secreted EVs by transferring miR-146a-5p in cervical cancer. After characterization of miR-146a-5p expression in clinical cervical cancer tissue samples, gain-
Tomofumi Yamamoto et al.
Blood advances, 6(8), 2480-2495 (2022-01-04)
Multiple myeloma (MM) is a hematopoietic malignancy whose prognosis has improved with the development of new agents such as lenalidomide over the last decade. However, long-term exposure to drugs induces the acquisition of resistance by MM cells and leads to treatment
Ana Lonic et al.
STAR protocols, 3(2), 101305-101305 (2022-05-03)
Previously published protocols for quantification of endosomal recycling are limited by the use of radioactive reagents, washing of cells in reducing buffers, or the requirement for large numbers of cells. Here, we describe a protocol for quantification of endosomal recycling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service