Skip to Content
Merck
All Photos(1)

Key Documents

F9054

Sigma-Aldrich

Fumitremorgin C

from Neosartorya fischeri, film

Synonym(s):

FTC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25N3O3
CAS Number:
Molecular Weight:
379.45
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

biological source

Neosartorya fischeri

Quality Level

form

film

solubility

acetonitrile: soluble 5 mg/mL
methanol: soluble 5 mg/mL
DMSO: soluble
chloroform: soluble

antibiotic activity spectrum

fungi

shipped in

wet ice

storage temp.

2-8°C

SMILES string

COc1ccc2c3C[C@@H]4N([C@@H](\C=C(/C)C)c3[nH]c2c1)C(=O)[C@@H]5CCCN5C4=O

InChI

1S/C22H25N3O3/c1-12(2)9-18-20-15(14-7-6-13(28-3)10-16(14)23-20)11-19-21(26)24-8-4-5-17(24)22(27)25(18)19/h6-7,9-10,17-19,23H,4-5,8,11H2,1-3H3/t17-,18-,19-/m0/s1

InChI key

DBEYVIGIPJSTOR-FHWLQOOXSA-N

General description

Chemical structure: indol derivative

Application

Fumitremorgin C has been used as adenosine triphosphate (ATP)-binding cassette superfamily G member 2 (ABCG2) inhibitor to study its effects on the cell viability of mouse neural stem/progenitor cells (NSPCs). It has also been used as an ABCG2 inhibitor to study its effects on lung cancer cells.

Biochem/physiol Actions

Fumitremorgin C (FTC) is a fungal toxin of the diketopiperazines family of compounds. In mammalian cells, FTC is tremorgenic and causes cell cycle arrest. Fumitremorgin C has been shown to reverse resistance to doxorubicin, mitoxantrane, and topotecan in non-Pgp (P-glyco­protein), non-MRP (multidrug resistance protein) multidrug-resistance (MDR) cells. This reversal of resistance is associated with an increase in drug accumulation. Fumitremorgin C is a specific, selective, and potent inhibitor at micromolar concentrations of the breast cancer resistant protein (BCRP/ABCG2), an ABC transporter associated with chemotherapy resistance. FTC, in combination with mitoxantrone, can be used for the detection of ABCG2 functional activity in several cell lines.
Selective inhibitor for the breast cancer resistance protein.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Packaging

Store desiccated at 2-8 °C. Under these conditions, the product is stable for 2 years. A solution in DMSO is stable for 2 months at 2-8 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting-Chun Kuo et al.
Asian journal of surgery, 43(1), 339-346 (2019-06-12)
Preoperative differentiation of follicular thyroid carcinoma (FTC) from follicular adenoma (FA) remains an unsolved puzzle. Patients sometimes undergo unnecessary lobectomy for histology confirmation inevitably. In this retrospective study, we propose new gray-scale ultrasonographic (US) features that may help to differentiate
H P M Natukunda et al.
South African medical journal = Suid-Afrikaanse tydskrif vir geneeskunde, 107(11), 965-975 (2017-12-22)
Studies investigating symptoms associated with combination antiretroviral therapy (cART) use among adolescents in resource-limited settings are rare beyond clinical trials. Identifying adolescents at risk of non-adherence is imperative for HIV/AIDS programming and controlling the epidemic in this key population. To
Ross D Cranston et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 64(5), 614-620 (2016-12-18)
Human immunodeficiency virus (HIV) disproportionately affects men who have sex with men (MSM) and transgender women (TGW). Safe and acceptable topical HIV prevention methods that target the rectum are needed. MTN-017 was a phase 2, 3-period, randomized sequence, open-label, expanded
Hans Minderman et al.
Cytometry, 48(2), 59-65 (2002-07-13)
The breast cancer resistance protein (BCRP) is an ATP-binding cassette (ABC) half-transporter that mediates energy-dependent drug efflux. Assessing the clinical relevance of the BCRP will require sensitive and specific methods for detecting its expression and function that allow high-volume specimen
Roy M Gulick et al.
The Journal of infectious diseases, 215(2), 238-246 (2016-11-05)
Maraviroc (MVC) is a candidate for human immunodeficiency virus (HIV) pre-exposure prophylaxis. Phase 2 48-week safety/tolerability study was conducted, comparing 4 regimens: MVC alone, MVC plus emtricitabine (FTC), MVC plus tenofovir disoproxil fumarate (TDF), and TDF plus FTC. Eligible participants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service