Skip to Content
Merck
All Photos(3)

Key Documents

F8257

Sigma-Aldrich

Flunarizine dihydrochloride

≥98% (TLC)

Synonym(s):

1-[Bis(4-fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)piperazine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26F2N2 · 2HCl
CAS Number:
Molecular Weight:
477.42
Beilstein:
4284243
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

originator

Johnson & Johnson

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Fc1ccc(cc1)C(N2CCN(CC2)C\C=C\c3ccccc3)c4ccc(F)cc4

InChI

1S/C26H26F2N2.2ClH/c27-24-12-8-22(9-13-24)26(23-10-14-25(28)15-11-23)30-19-17-29(18-20-30)16-4-7-21-5-2-1-3-6-21;;/h1-15,26H,16-20H2;2*1H/b7-4+;;

InChI key

RXKMOPXNWTYEHI-RDRKJGRWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flunarizine has antihistamine, anticonvulsant and vestibular depressive properties. It protects endothelial cells from calcium overload-induced damage. Flunarizine is used to treat epilepsy, migraine prophylaxis, cerebral blood flow disturbances, peripheral vascular disease and vertigo.

Application

Flunarizine dihydrochloride has been used to study the effect of flunarizine on epileptiform activity. It is also used to study its effect on high-potassium induced increase of intracellular free calcium.

Biochem/physiol Actions

Blocks T-type Ca2+/Na+ channels.
Blocks T-type Ca2+/Na+ channels; inhibits K+-induced catecholamine release from chromaffin cells

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of flunarizine on induced calcium transients as measured in fura-2-loaded neurons of the rat dorsal root ganglion
Leybaert L, et al.
Naunyn-Schmiedeberg'S Archives of Pharmacology, 348(3), 269-274 (1993)
Double-blind placebo-controlled trial of flunarizine as add-on therapy in epilepsy
Overweg J, et al.
Epilepsia, 25(2), 217-222 (1984)
Flunarizine
Holmes B, et al.
Drugs, 27(1), 6-44 (1984)
Barbara Budzynska et al.
Behavioural brain research, 228(1), 144-150 (2011-12-20)
The motivational component of drug withdrawal may contribute to drug seeking and relapse through the negative reinforcement-related process; thus, it is important to understand the mechanisms that mediate affective withdrawal behaviors. The present study was undertaken to examine the calcium-dependent
The effect of flunarizine on penicillin model epilepsy in rats
BAGIRICI F, et al.
Journal of Experimental and Clinical Medicine, 16(4) (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service