Skip to Content
Merck
All Photos(1)

Key Documents

A-911

Supelco

7-Aminoflunitrazepam solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14FN3O
CAS Number:
Molecular Weight:
283.30
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

FC1=CC=CC=C1C2=NCC(N(C)C3=CC=C(N)C=C23)=O

InChI

1S/C16H14FN3O/c1-20-14-7-6-10(18)8-12(14)16(19-9-15(20)21)11-4-2-3-5-13(11)17/h2-8H,9,18H2,1H3

InChI key

LTCDLGUFORGHGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

As a major metabolite of flunitrazepam, 7-aminoflunitrazepam is a biomarker for use of the parent drug. Flunitrazepam is an intermediate-acting benzodiazepine, typically used in the treatment of severe insomnia. This certified solution standard is suitable for numerous GC/MS or LC/MS testing applications in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yi-Chi Ho et al.
Analytica chimica acta, 697(1-2), 1-7 (2011-06-07)
Despite the advantages of simplicity and high-throughput detection that matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS) has over other methods, quantitative analysis of low-molecular-weight analyte is hampered by interference from matrix-derived background noise and signal fluctuation due to the
Franz E Dussy et al.
International journal of legal medicine, 120(6), 323-330 (2005-10-13)
A high-performance liquid chromatography method for the determination of benzodiazepines and their metabolites in whole blood and serum using mass spectrometry (MS) and photodiode array (PDA) detection is presented. The combination of both detection types can complement each other and
Chiung-Wei Huang et al.
Journal of chromatography. A, 1110(1-2), 240-244 (2006-02-18)
A sweeping technique, in conjunction with micellar electrokinetic chromatography, for the simultaneous determination of flunitrazepam and its major metabolites, 7-aminoflunitrazepam and N-desmethylflunitrazepam, is described. The optimized conditions for the sweeping and separation were a pH 9.5 buffer, 25mM borate, 50mM
H L Lord et al.
Journal of pharmaceutical and biomedical analysis, 40(3), 769-780 (2005-12-15)
We report on the development of solid phase microextraction probes for drug analysis, prepared with antibodies specific for benzodiazepines covalently immobilized to the surface. In the technique, immobilized antibody probes are exposed to a sample containing the drug for 30
I Gafni et al.
Journal of clinical psychopharmacology, 23(2), 169-175 (2003-03-18)
Flunitrazepam, a hypnotic benzodiazepine, is widely prescribed around the world for the treatment of insomnia and as a preanesthetic. In vitro studies have shown that the metabolism of flunitrazepam to desmethylflunitrazepam and 3-hydroxyflunitrazepam is mediated in part by the polymorphic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service