F8016
Ficoll® 400
lyophilized powder, Type 400-DL, BioReagent, suitable for cell culture
Synonym(s):
Polysucrose 400
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ficoll® 400 has been used for the isolation of mesenchymal stem cells from rat bone marrow and preparation of Ficoll cushion solution .
Used for cell separation and organelle isolation.
Other Notes
A nonionic synthetic polymer of sucrose.
Dialyzed
Legal Information
Ficoll is a registered trademark of Cytiva
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Effect of bone marrow versus adipose tissue derived mesenchymal stem cells on the pancreas of streptozotocin-induced diabetes mellitus type I in adult male rats (Histological Study)
Omar AI, et al.
The Egyptian journal of histology., 40(1), 12-24 (2017)
Tianjin Liu et al.
Experimental & molecular medicine, 49(6), e350-e350 (2017-07-01)
α-cells, which synthesize glucagon, also support β-cell survival and have the capacity to transdifferentiate into β-cells. However, the role of α-cells in pathological conditions and their putative clinical applications remain elusive due in large part to the lack of mature
Isolation of centrosomes from cultured Mammalian cells
Meigs TE and Kaplan DD
Cold Spring Harbor Protocols, 2008(8), pdb-prot5039 (2008)
Jyoti Pant et al.
PLoS pathogens, 17(9), e1008768-e1008768 (2021-09-25)
Trypanosome Lytic Factor (TLF) is a primate-specific high-density lipoprotein (HDL) complex that, through the cation channel-forming protein apolipoprotein L-1 (APOL1), provides innate immunity to select kinetoplastid parasites. The immunoprotective effects of TLF have been extensively investigated in the context of
Effect of bone marrow derived mesenchymal stem cells on healing of induced full-thickness skin wounds in albino rat.
Basiouny HS, et al.
International journal of stem cells, 6(1), 12-25 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service