Skip to Content
Merck
All Photos(2)

Key Documents

F5137

Sigma-Aldrich

Anti-Sheep IgG (whole molecule)−FITC antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Rabbit Anti-Sheep IgG (whole molecule)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

FITC conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100
indirect immunofluorescence: 1:100

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. Anti-Sheep IgG (whole molecule)-FITC antibody is specific for sheep IgG. Rabbit Anti-Sheep IgG is conjugated to Fluorescein Isothiocyanate (FITC) in an alkaline reaction, then further purified to remove unbound FITC.

Immunogen

Purified sheep IgG

Application

Anti-Sheep IgG (whole molecule) FITC antibody produced in rabbit has been used in indirect immunofluorescence assay (IFAT) and immunofluorescence staining.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yanina P Hecker et al.
Tropical animal health and production, 45(7), 1645-1647 (2013-03-16)
The aim of this study was to describe the occurrence of antibodies to Toxoplasma gondii and Neospora caninum in dairy sheep from the Humid Pampa region, Argentina. Blood samples from 704 dairy sheep belonging to six flocks were collected. Using
Larissa Martins de Brito Moraes et al.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria, 20(4), 312-317 (2011-12-15)
Neosporosis and toxoplasmosis are parasitic diseases which can cause reproductive problems in goats and sheep. The current study aimed to determine the occurrence of anti-Neospora caninum and anti-Toxoplasma gondii IgG antibodies in goats and sheep from the districts of Amarante
Joanna-Marie Howes et al.
Journal of thrombosis and haemostasis : JTH, 18(4), 942-954 (2020-01-03)
Atherosclerotic plaque rupture and subsequent thrombosis underpin thrombotic syndromes. Under inflammatory conditions in the unstable plaque, perturbed endothelial cells secrete von Willebrand Factor (VWF) which, via its interaction with GpIbα, enables platelet rolling across and adherence to the damaged endothelium.
Occurrences of anti-Toxoplasma gondii and anti-Neospora caninum antibodies in sheep from four districts of Tocantins state, Brazilian Legal Amazon Region
Guimaraes A, et al.
Pesquisa Veterinária Brasileira, 35(2), 110-114 (2015)
Yi Wen et al.
American journal of physiology. Renal physiology, 318(1), F107-F116 (2019-11-19)
Nephrotoxic serum nephritis (NTN) models immune-mediated human glomerulonephritis and culminates in kidney inflammation and fibrosis, a process regulated by T lymphocytes. TNF-α is a key proinflammatory cytokine that contributes to diverse forms of renal injury. Therefore, we posited that TNF-α

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service