Skip to Content
Merck
All Photos(2)

Key Documents

F7512

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)−FITC antibody produced in sheep

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Sheep Anti-Rabbit IgG (whole molecule)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

sheep

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:160
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:160

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Rabbit IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in rabbit serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.
Fluorescein isothiocyanate (FITC) is a fluorescein derivative (fluorochrome) used to tag antibodies, including secondary antibodies, for use in fluorescence-based assays and procedures. FITC excites at 495 nm and emits at 521 nm.
Sheep polyclonal anti-Rabbit IgG (whole molecule)−FITC antibody binds normal rabbit serum and rabbit IgG. It does not cross react with human IgG.

Application

Anti-Rabbit IgG (whole molecule)-FITC antibody produced in sheep has been used in:
  • immunocytochemistry
  • immunofluorescence
  • immunohistochemistry

Sheep polyclonal anti-Rabbit IgG (whole molecule)−FITC antibody may be used to detect and quantitate the level of IgG in rabbit serum and biological fluids by fluorescent techniques.

Other Notes

Antibody adsorbed with human IgG.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erika Cecon et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(6), 2566-2582 (2015-03-12)
Melatonin is the hormone produced by the pineal gland known to regulate physiologic rhythms and to display immunomodulatory and neuroprotective properties. It has been reported that Alzheimer disease patients show impaired melatonin production and altered expression of the 2 G
Involvement of high mobility group box 1 in the development and maintenance of chemotherapy-induced peripheral neuropathy in rats
Nishida T, et al.
Toxicology, 89, 14-24 (2016)
Matrix stiffness modulates formation and activity of neuronal networks of controlled architectures
Lantoine J, et al.
Biomaterials, 1863(4), 673-685 (2016)
Sandra Marcia Muxel et al.
Frontiers in immunology, 9, 2792-2792 (2018-12-18)
Parasite recognition by Toll-like receptors (TLRs) contributes to macrophage activation and subsequent control of Leishmania infection through the coordinated production of pro-inflammatory and microbicidal effector molecules. The modulation of microRNA (miRNA) expression by Leishmania infection potentially mediates the post-transcriptional regulation
Xiaoli He et al.
Journal of molecular cell biology, 2(4), 199-208 (2010-06-30)
High renewal and maintenance of multipotency of human adult stem cells (hSCs), are a prerequisite for experimental analysis as well as for potential clinical usages. The most widely used strategy for hSC culture and proliferation is using serum. However, serum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service