Skip to Content
Merck

Skip To

F4767

Filipin III from Streptomyces filipinensis

≥85% (HPLC)

Synonym(s):

Filimarisin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C35H58O11
CAS Number:
Molecular Weight:
654.83
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥85% (HPLC)

form

powder

mp

163-180  °C

solubility

methanol: 9.80-10.20 mg/mL, clear, colorless to yellow

antibiotic activity spectrum

fungi, yeast

mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

CCCCCC(O)C1C(O)CC(O)CC(O)CC(O)CC(O)CC(O)CC(O)C(C)=C\C=C\C=C\C=C\C=C\C(O)C(C)OC1=O

InChI

1S/C35H58O11/c1-4-5-11-16-31(42)34-33(44)22-29(40)20-27(38)18-25(36)17-26(37)19-28(39)21-32(43)23(2)14-12-9-7-6-8-10-13-15-30(41)24(3)46-35(34)45/h6-10,12-15,24-34,36-44H,4-5,11,16-22H2,1-3H3/b7-6+,10-8+,12-9+,15-13+,23-14-

InChI key

IMQSIXYSKPIGPD-BWJOQPJXSA-N

Application

Filipin has been used in a double staining procedure as a probe for the detection of lipoproteins in polyacrylamide gel and immobilized on nitrocellulose membranes.
Filipin has been used in a double staining procedure as a probe for the detection of lipoproteins in polyacrylamide gel and immobilized on nitrocellulose membranes. It is also widely used to localize and quantitate unesterified cholesterol by virtue of a specific fluorescent complex.

Biochem/physiol Actions

Filipin is a polyene macrolide antibiotic and antifungal. The antifungal mechanism of action may be due to altering membrane permeability and associated functions by binding to membrane sterols. Filipin inhibits prion protein (PrP) endocytosis and causes the release of PrP from the plasma membrane.

Other Notes

Filipin III is the major component of 8 isomeric components in filipin.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F9765M2147M6418
assay

≥85% (HPLC)

assay

≥70% (UV)

assay

≥98% (HPLC)

assay

≥97% (HPLC)

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

200

form

powder

form

powder

form

crystalline powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

methanol: 9.80-10.20 mg/mL, clear, colorless to yellow

solubility

methanol: 9.80-10.20 mg/mL, clear to hazy, yellow to very deep greenish-yellow

solubility

ethanol: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

solubility

methanol: 9.80-10.20 mg/mL, clear, yellow to very deep yellow

mp

163-180  °C

mp

-

mp

175.4 °C

mp

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet


Dimitri Moreau et al.
EMBO reports, 20(7), e47055-e47055 (2019-07-04)
Most cells acquire cholesterol by endocytosis of circulating low-density lipoproteins (LDLs). After cholesteryl ester de-esterification in endosomes, free cholesterol is redistributed to intracellular membranes via unclear mechanisms. Our previous work suggested that the unconventional phospholipid lysobisphosphatidic acid (LBPA) may play
T N Efremova et al.
Tsitologiia, 54(6), 508-514 (2012-09-25)
Cholesterol is one of the major lipid components of plasma membrane and it plays an important role in various signaling processes in mammalian cells. Our study focused on the role of membrane cholesterol in organization and dynamics of actin cytoskeleton.
Henrik Tjellström et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(4), 1128-1138 (2009-12-08)
As in other eukaryotes, plant plasma membranes contain sphingolipids, phospholipids, and free sterols. In addition, plant plasma membranes also contain sterol derivatives and usually <5 mol% of a galactolipid, digalactosyldiacylglycerol (DGDG). We earlier reported that compared to fully fertilized oats



Global Trade Item Number

SKUGTIN
F4767-1MG04061838446060
F4767-5MG04061832947563

Questions

  1. Good afternoon; I would be very grateful if you could clarify me which is the best solvent for filipin III (F4767) stock solution preparation. Product specification is different from product information (methanol; ethanol; DMSO..). Thank you in advance.

    1 answer
    1. As provided, every batch of this product released has been tested for solubility in methanol at concentrations up to about 10 mg/mL. It is also known to be soluble in organic solvents, e.g., ethanol (2 mg/mL), DMSO (5 mg/mL), and dimethyl formamide (10 mg/mL). The optimal solvent may vary depending on the specific application.

      Helpful?

Reviews

No rating value

Active Filters