Skip to Content
Merck
All Photos(6)

Documents

F3381

Sigma-Aldrich

Fast Red Violet LB Salt

Dye content ≥90%, Powder

Synonym(s):

5-Chloro-4-benzamido-2-methylbenzenediazonium chloride hemi(zinc chloride) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H11Cl2N3O · ½ZnCl2
CAS Number:
Molecular Weight:
376.31
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Fast Red Violet LB Salt, Dye content ≥90 %

form

powder

composition

Dye content, ≥90%

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Cc1cc(NC(=O)c2ccccc2)c(Cl)cc1[N+]#N

InChI

1S/C14H10ClN3O.ClH/c1-9-7-13(11(15)8-12(9)18-16)17-14(19)10-5-3-2-4-6-10;/h2-8H,1H3;1H

InChI key

SARKXLKWFKNUMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fast Red Violet LB Salt, also known as 4′-amino-2′-chloro-5′-methylbenzanilide. It is a lipophilic amine with an aromatic system of moderate size and a hydrophilic diazonium cation. The diazonium salt is available commercially as a zinc chloride double salt. It comprises ≥90% dye content.

Application

  • Fast Red Violet LB Salt is suitable for quantitating alkaline phosphatase through histochemical, hematological, histological, and flow cytometric methods.
  • It has been used to study the role of TNF-α during fracture healing.
  • has been used for the detection of tartrate-resistant acid phosphatase (TRAP) in tissue sections.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impaired fracture healing in the absence of TNF-alpha signaling: the role of TNF-alpha in endochondral cartilage resorption
Gerstenfeld, et al.
Journal of Bone and Mineral Research, 18(9), 1584?1592-1584?1592 (2003)
Three-dimensional reconstruction of fracture callus morphogenesis.
Gerstenfeld LC
The Journal of Histochemistry and Cytochemistry, 54, 1215-1215 (2006)
Tatsuya Yoshida et al.
Journal of orthopaedic surgery and research, 4, 31-31 (2009-08-05)
Pasteurized bone grafting is used following the excision of a bone tumor for the purpose of eliminating neoplastic cells while preserving bone-inducing ability. In the hopes of guaranteeing the most favourable results, the establishment of an animal model has been
Conn's Biological Stains
Kiernan & Horobin
Conn?s Biological Stains (2002)
Comparison of effects of the bisphosphonate alendronate versus the RANKL inhibitor denosumab on murine fracture healing.
Gerstenfeld LC
Journal of Bone and Mineral Research, 24, 196-196 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service