Skip to Content
Merck
All Photos(1)

Documents

F4648

Sigma-Aldrich

Fast Red TR/Naphthol AS-MX

SIGMAFAST, fluorogenic, tablet

Synonym(s):

4-Chloro-2-methylbenzenediazonium / 3-Hydroxy-2-naphthoic acid 2,4-dimethylanilide phosphate tablets

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

product name

SIGMAFAST Fast Red TR/Naphthol AS-MX Tablets, tablet, To prepare 1 mL

form

tablet

solubility

water: 1 tablet/mL, clear to hazy, colorless to faintly yellow

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

SIGMAFAST Fast Red TR/Naphthol AS-MX Tablets has been used:
  • in alkaline phosphatase staining in organ sections
  • for staining chondrogenic spheroids in immunohistochemistry studies
  • for the detection of hybridization signals in in situ mRNA hybridization studies of human palatine tonsils sections

Developed for use as a precipitating substrate for the detection of alkaline phosphatase activity in immunochemistry. Primary uses are in immunohistology, immunoblotting, or dot blotting. SIGMAFAST Fast Red TR/Naphthol AS-MX tablets contain 0.6 mM levamisole to block endogenous alkaline phosphatase activity. SIGMAFAST Fast Red TR/Naphthol AS-MX Tablets require no additional steps or buffers to prepare an active substrate solution.

Reconstitution

Each tablet dissolved in 1 mL deionized water yields a ready-to-use buffered solution.

Legal Information

SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn B Moore et al.
Developmental biology, 437(1), 27-40 (2018-03-09)
Neural basic helix-loop helix (bHLH) transcription factors promote progenitor cell differentiation by activation of downstream target genes that coordinate neuronal differentiation. Here we characterize a neural bHLH target gene in Xenopus laevis, vexin (vxn; previously sbt1), that is homologous to
Anja Rabien et al.
International journal of molecular sciences, 19(6) (2018-06-03)
Bladder cancer still requires improvements in diagnosis and prognosis, because many of the cases will recur and/or metastasize with bad outcomes. Despite ongoing research on bladder biomarkers, the clinicopathological impact and diagnostic function of miRNA maturation regulators Drosha and Argonaute
Jason S Williams et al.
Developmental biology, 444 Suppl 1, S274-S286 (2018-04-01)
Melanocytes derive from neural crest cells, which are a highly migratory population of cells that play an important role in pigmentation of the skin and epidermal appendages. In most vertebrates, melanocyte precursor cells migrate solely along the dorsolateral pathway to
Tony Y-C Tsai et al.
Science (New York, N.Y.), 370(6512), 113-116 (2020-10-03)
Animal development entails the organization of specific cell types in space and time, and spatial patterns must form in a robust manner. In the zebrafish spinal cord, neural progenitors form stereotypic patterns despite noisy morphogen signaling and large-scale cellular rearrangements
Telomere length and telomerase activity during expansion and differentiation of human mesenchymal stem cells and chondrocytes
Parsch D, et al.
Journal of Molecular Medicine, 82(1), 49-55 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service