Skip to Content
Merck
All Photos(1)

Key Documents

F3055

Sigma-Aldrich

Flavopiridol hydrochloride hydrate

≥98% (HPLC), powder

Synonym(s):

(−)-2-(2-Chlorophenyl)-5,7-dihydroxy-8-[(3S,4R)-3-hydroxy-1-methyl-4-piperidinyl]-4H-1-benzopyran-4-one hydrochloride, L-86-8276, NSC-649890

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20ClNO5 ·HCl · xH2O
CAS Number:
Molecular Weight:
438.30 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

H2O: ~2 mg/mL
DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

OC1=C(C(C=C(C2=C(Cl)C=CC=C2)O3)=O)C3=C([C@H]4CCN(C)C[C@H]4O)C(O)=C1.Cl

InChI

1S/C21H20ClNO5.ClH/c1-23-7-6-12(17(27)10-23)19-14(24)8-15(25)20-16(26)9-18(28-21(19)20)11-4-2-3-5-13(11)22;/h2-5,8-9,12,17,24-25,27H,6-7,10H2,1H3;1H/t12-,17+;/m0./s1

InChI key

LGMSNQNWOCSPIK-LWHGMNCYSA-N

Application

Flavopiridol hydrochloride hydrate has been used:
  • as a cyclin-dependent kinase 9 (CDK9) inhibitor to study its effects on histone H3 methylation at lysine 36 (H3K36) and deactivation of transcription in porcine fetal fibroblasts
  • as an RNA polymerase inhibitor to study its effects on hepatic cells
  • as RNA transcription inhibitor to study its effects on euchromatin coarsening in zebrafish embryo

Biochem/physiol Actions

Flavopiridol hydrochloride is a potent CDK (cyclin-dependent kinase), and a CDC25 phosphatase family inhibitor.
Flavopiridol is a semi-synthetic flavone obtained from Dysoxylum binectariferum that acts as an anti-tumor agent against several cancers. It also shows anti-cancer properties due to which it has been studied in the treatment of acute myeloid leukemia (AML).

Features and Benefits

This compound is featured on the CDKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saowaluk Saisomboon et al.
Heliyon, 5(5), e01675-e01675 (2019-06-14)
Flavopiridol, a pan-cyclin-dependent kinase (CDK) inhibitor, was recently identified as an effective antitumor agent for several cancers. We investigated the antitumor effect of flavopiridol on cholangiocarcinoma (CCA), in vitro and in vivo. A methylthiotetrazole assay revealed that the proliferation of
Uma Raju et al.
Cancer research, 63(12), 3263-3267 (2003-06-18)
Flavopiridol, a cyclin-dependent kinase (cdk) inhibitor, can cause cell cycle arrest, induce apoptosis in cancer cells, and inhibit tumor cell growth in vivo. The present study investigated the in vitro radiosensitizing effect of flavopiridol and the underlying molecular mechanisms in
Elisa Barbieri et al.
Cell reports, 33(6), 108373-108373 (2020-11-12)
Genome-wide profiling of nascent RNA has become a fundamental tool to study transcription regulation. Unlike steady-state RNA-sequencing (RNA-seq), nascent RNA profiling mirrors real-time activity of RNA polymerases and provides an accurate readout of transcriptome-wide variations. Some species of nuclear RNAs
Stefano de Pretis et al.
Genome research, 27(10), 1658-1664 (2017-09-15)
Overexpression of the MYC transcription factor causes its widespread interaction with regulatory elements in the genome but leads to the up- and down-regulation of discrete sets of genes. The molecular determinants of these selective transcriptional responses remain elusive. Here, we
Anne Pumfery et al.
Current pharmaceutical design, 12(16), 1949-1961 (2006-06-22)
Cyclin-dependent kinases (CDKs) are key regulators of the cell cycle and RNA polymerase II transcription. Several pharmacological CDK inhibitors (PCIs) are currently in clinical trials as potential cancer therapeutics since CDK hyperactivation is detected in the majority of neoplasias. Within

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service