Skip to Content
Merck

Skip To

F1502

D-Fructose 6-phosphate dipotassium salt

≥97% (enzymatic), amorphous powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C6H11O9PK2
CAS Number:
Molecular Weight:
336.32
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (inorganic)

Quality Level

assay

≥97% (enzymatic)

form

amorphous powder

impurities

≤0.05 mol % fructose 1,6-diphosphate, ≤1.5 mol % glucose 6-phosphate

color

white to off-white

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

cation traces

K: 20.3-26.2% (dry basis)

storage temp.

−20°C

SMILES string

[K+].[K+].OCC1(O)O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@@H]1O

InChI

1S/C6H13O9P.2K/c7-2-6(10)5(9)4(8)3(15-6)1-14-16(11,12)13;;/h3-5,7-10H,1-2H2,(H2,11,12,13);;/q;2*+1/p-2/t3-,4-,5+,6?;;/m1../s1

InChI key

UNAFCPBHNZYEIY-UHOQFTJOSA-L

Application

D-Fructose 6-phosphate (F6P) is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amino-transferase(s) and glucosamine-6P synthase(s).[1]

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F3627D3126P7877
biological source

synthetic (inorganic)

biological source

bacterial (Corynebacterium)

biological source

synthetic (inorganic)

biological source

synthetic (organic)

assay

≥97% (enzymatic)

assay

≥98%

assay

≥95% (ICP-AES)

assay

≥97% (enzymatic)

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

solubility

H2O: 100 mg/mL, clear, colorless to very faintly yellow

solubility

water: 50 mg/mL, clear, colorless

form

amorphous powder

form

amorphous powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Joel L Asenjo et al.
Biochimica et biophysica acta, 1840(6), 1798-1807 (2014-01-22)
Fructose-1,6-bisphosphatase, a major enzyme of gluconeogenesis, is inhibited by AMP, Fru-2,6-P2 and by high concentrations of its substrate Fru-1,6-P2. The mechanism that produces substrate inhibition continues to be obscure. Four types of experiments were used to shed light on this:
Chia-I Lin et al.
Journal of the American Chemical Society, 136(3), 906-909 (2014-01-02)
Lincomycin A is a clinically useful antibiotic isolated from Streptomyces lincolnensis. It contains an unusual methylmercapto-substituted octose, methylthiolincosamide (MTL). While it has been demonstrated that the C8 backbone of MTL moiety is derived from D-fructose 6-phosphate and D-ribose 5-phosphate via
Yimin Qian et al.
Bioorganic & medicinal chemistry letters, 21(21), 6264-6269 (2011-10-01)
Through high throughput screening and subsequent hit identification and optimization, we synthesized a series of 1-arylcarbonyl-6,7-dimethoxyisoquinoline derivatives as the first reported potent and reversible GFAT inhibitors. SAR studies of this class of compounds indicated significant impact on GFAT inhibition potency



Global Trade Item Number

SKUGTIN
F1502-1G04061833611975
F1502-500MG04061833611982

Questions

Reviews

No rating value

Active Filters