Documents
EP5424704000
Eppendorf® Rotor F-45-32-5-PCR
incl. lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 4 x PCR strips
incl. lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5424704000
parameter
15,000 max. RCF (18,615 xg)
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with centrifuge models 5424/5424R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG