Skip to Content
Merck
All Photos(1)

Key Documents

EHU033071

Sigma-Aldrich

MISSION® esiRNA

targeting human CDK6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGTTTCAGCTTCTCCGAGGTCTGGACTTTCTTCATTCACACCGAGTAGTGCATCGCGATCTAAAACCACAGAACATTCTGGTGACCAGCAGCGGACAAATAAAACTCGCTGACTTCGGCCTTGCCCGCATCTATAGTTTCCAGATGGCTCTAACCTCAGTGGTCGTCACGCTGTGGTACAGAGCACCCGAAGTCTTGCTCCAGTCCAGCTACGCCACCCCCGTGGATCTCTGGAGTGTTGGCTGCATATTTGCAGAAATGTTTCGTAGAAAGCCTCTTTTTCGTGGAAGTTCAGATGTTGATCAACTAGGAAAAATCTTGGACGTGATTGGACTCCCAGGAGAAGAAGACTGGCCTAGAGATGTTGCCCTTCCCAGGCAGGCTTTTCATTCAAAATCTGCCCAACCAATTGAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Gong et al.
Aging, 12(12), 12086-12106 (2020-06-26)
Emerging studies indicate that long non-coding RNAs (lncRNAs) play crucial roles in colorectal cancer (CRC). Here, we reported lncRNA CASC21, which is induced by FOXP1, functions as an oncogene in CRC. We systematically elucidated its clinical significance and possible molecular
Lihua Guo et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 23(3), 787-794 (2018-07-14)
Clear cell renal cell carcinoma (ccRCC) accounts for 70-80% of renal cell carcinomas. Various microRNAs (miRs) have been reported to affect the tumorigenesis of ccRCC. However, the role of miR-384 in ccRCC is still unknown. Thus, the purpose of this
Míriam Tarrado-Castellarnau et al.
Molecular systems biology, 13(10), 940-940 (2017-10-06)
Cyclin-dependent kinases (CDK) are rational cancer therapeutic targets fraught with the development of acquired resistance by tumor cells. Through metabolic and transcriptomic analyses, we show that the inhibition of CDK4/6 leads to a metabolic reprogramming associated with gene networks orchestrated
Xuefeng Pan et al.
Molecular therapy. Nucleic acids, 22, 38-49 (2020-09-11)
Emerging studies indicate that long noncoding RNAs (lncRNAs) play crucial roles in ovarian cancer (OC). By analyzing high-throughput data, we found that SNHG17 was highly expressed in multiple OC cohorts. However, its functions in OC were not explored. In this
Tongzheng Liu et al.
Nature communications, 8, 13923-13923 (2017-01-10)
Tumour metastasis, the spread of cancer cells from the original tumour site followed by growth of secondary tumours at distant organs, is the primary cause of cancer-related deaths and remains poorly understood. Here we demonstrate that inhibition of CDK4/6 blocks

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service