Skip to Content
Merck
All Photos(1)

Key Documents

E8534

Sigma-Aldrich

E3330

≥98% (HPLC)

Synonym(s):

(2E)-2-[(4,5-Dimethoxy-2-methyl-3,6-dioxo-1,4-cyclohexadien-1-yl)methylene]-undecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O6
CAS Number:
Molecular Weight:
378.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

red to orange

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

CCCCCCCCC\C(=C/C1=C(C)C(=O)C(OC)=C(OC)C1=O)C(O)=O

InChI

1S/C21H30O6/c1-5-6-7-8-9-10-11-12-15(21(24)25)13-16-14(2)17(22)19(26-3)20(27-4)18(16)23/h13H,5-12H2,1-4H3,(H,24,25)/b15-13+

InChI key

AALSSIXXBDPENJ-FYWRMAATSA-N

Application

E3330 has been used in the inhibition of endothelial cancer cells and apurinic/apyrimidinic endodeoxyribonuclease 1 (APEX1).

Biochem/physiol Actions

E3330 is a specific inhibitor of AP endonuclease 1 redox domain. E3330 inhibits APE-1 regulation of transcription factors, but does not affect Ape1 DNA repair activity. AP endonuclease 1 (APE1; also known as REF-1) is a multifunctional protein with dual functions in DNA repair and redox regulation of transcription factors. It is involved in apurinic/apyrimidinic endonuclease DNA base excision repair activity, in proofreading exonuclease activity, and in modulating DNA binding activity of several transcription factors including NF-κB, Egr-1, p53, AP-1, CREB, HIF-α, and members of the Pax family. APE1 is overexpressed in several human cancers, and disruption of APE1 function has detrimental effects on cancer cell viability. E3330 significantly reduces the growth of human pancreatic cancer cells in vitro and inhibits pancreatic cancer cell migration.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Shimizu et al.
Nature biotechnology, 18(8), 877-881 (2000-08-10)
We have developed a method using novel latex beads for rapid identification of drug receptors using affinity purification. Composed of a glycidylmethacrylate (GMA) and styrene copolymer core with a GMA polymer surface, the beads minimize nonspecific protein binding and maximize
[Elucidation of pharmacological mechanism of drugs by studying the receptors].
Hiroshi Handa
The Japanese journal of antibiotics, 56 Suppl A, 39-46 (2003-12-19)
Andrej Jedinak et al.
Anticancer research, 31(2), 379-385 (2011-03-08)
The multi-functional apyrimidinic endonuclease 1/redox factor-1 (APE1/Ref-1) DNA repair and redox signaling protein has been shown to have a role in cancer growth and survival, however, little has been investigated concerning its role in inflammation. In this study, an APE1
Yukiko Saitou et al.
World journal of gastroenterology, 11(40), 6258-6261 (2006-01-19)
To investigate the reduction of cell viability in human hepatocellular carcinoma (HCC) cell lines induced by inhibition of nuclear factor kappa B (NF kappa B). HLE, SKHep1, and HepG2 were incubated and E3330 was used to compare the stimulation of
Brittney A Manvilla et al.
Biochemistry, 50(48), 10540-10549 (2011-10-29)
AP endonuclease 1 (APE1) is a multifaceted protein with essential roles in DNA repair and transcriptional regulation. APE1 (ref-1) activates many transcription factors (TF), including AP-1 and NF-κB. While the mechanism of APE1 redox activity remains unknown, it may involve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service