Skip to Content
Merck
All Photos(1)

Key Documents

B5774

Supelco

Benzbromarone

analytical standard

Synonym(s):

3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethylbenzofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12Br2O3
CAS Number:
Molecular Weight:
424.08
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

~95%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CCc1oc2ccccc2c1C(=O)c3cc(Br)c(O)c(Br)c3

InChI

1S/C17H12Br2O3/c1-2-13-15(10-5-3-4-6-14(10)22-13)16(20)9-7-11(18)17(21)12(19)8-9/h3-8,21H,2H2,1H3

InChI key

WHQCHUCQKNIQEC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiqing Wu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 911, 122-132 (2012-12-12)
A high performance liquid chromatography-quadrupole time of flight mass spectrometry (HPLC-QTOF-MS) method was employed in investigation of benzbromarone metabolites in rat plasma, urine, feces and bile samples. Meanwhile, the metabolic pathways of benzbromarone in rats were discussed. The identification was
Kaoru Kobayashi et al.
Biopharmaceutics & drug disposition, 33(8), 466-473 (2012-08-31)
Benzbromarone (BBR) is metabolized to 1'-hydroxy BBR and 6-hydroxy BBR in the liver. 6-Hydroxy BBR is further metabolized to 5,6-dihydroxy BBR. The aim of this study was to identify the CYP isozymes involved in the metabolism of BBR to 1'-hydroxy
Ming-Han H Lee et al.
Drug safety, 31(8), 643-665 (2008-07-19)
Benzbromarone, a potent uricosuric drug, was introduced in the 1970s and was viewed as having few associated serious adverse reactions. It was registered in about 20 countries throughout Asia, South America and Europe. In 2003, the drug was withdrawn by
Zhijian Lin et al.
Journal of medicinal food, 17(11), 1214-1221 (2014-10-15)
Inulin, a group of dietary fibers, is reported to improve the metabolic disorders. In the present study, we investigated the effects of chicory inulin on serum metabolites of uric acid (UA), lipids, glucose, and abdominal fat deposition in quail model
Wenjing Zhu et al.
Life sciences, 91(11-12), 369-376 (2012-08-23)
Uric acid nephropathy (UAN) is due to excessive uric acid, which leads to hyperuricemia and kidney damage via the deposition of urate microcrystals in the kidneys. Iridoid glycosides of Paederia scandens (IGPS) is a major active component isolated from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service