Skip to Content
Merck
All Photos(1)

Key Documents

E5406

Sigma-Aldrich

Ethopropazine hydrochloride

≥98% (HPLC), powder

Synonym(s):

10-(2-Diethylaminopropyl)phenothiazine, 10-[2-(Diethylamino)propyl]phenothiazine hydrochloride, Dibutil hydrochloride, Parkin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2S · HCl
CAS Number:
Molecular Weight:
348.93
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

DMSO: >5 mg/mL at ~60 °C, clear

storage temp.

2-8°C

SMILES string

Cl[H].CCN(CC)C(C)CN1c2ccccc2Sc3ccccc13

InChI

1S/C19H24N2S.ClH/c1-4-20(5-2)15(3)14-21-16-10-6-8-12-18(16)22-19-13-9-7-11-17(19)21;/h6-13,15H,4-5,14H2,1-3H3;1H

InChI key

VXPCQISYVPFYRK-UHFFFAOYSA-N

Gene Information

human ... CHRM1(1128)

Looking for similar products? Visit Product Comparison Guide

General description

Ethopropazine hydrochloride possess anticholinergic, antihistamine and antiadrenergic properties.

Biochem/physiol Actions

Ethoproprazine hydrochloride is an inhibitor of butyrylcholinesterase; antiparkinsonian. It reduces extrapyramidal motor effects, characteristic of Parkinson′s disease; also alleviates thermal hyperalgesia in rats.

Features and Benefits

This compound is featured on the Acetylcholine Synthesis and Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickie C Chan et al.
Human molecular genetics, 20(9), 1726-1737 (2011-02-08)
Parkin, an E3 ubiquitin ligase implicated in Parkinson's disease, promotes degradation of dysfunctional mitochondria by autophagy. Using proteomic and cellular approaches, we show that upon translocation to mitochondria, Parkin activates the ubiquitin-proteasome system (UPS) for widespread degradation of outer membrane
Min Fu et al.
Molecular biology of the cell, 24(8), 1153-1162 (2013-02-22)
Glycoprotein 78 (Gp78) is a critical E3 ubiquitin ligase in endoplasmic reticulum-associated degradation. Overexpression of Flag-tagged Gp78 (Flag-gp78), but not Flag-gp78 mutated in its RING-finger domain (Flag-RINGmut) with deficient ubiquitin ligase activity, induces mitochondrial fragmentation and ubiquitination and proteasome-dependent degradation
Kei Okatsu et al.
Nature communications, 3, 1016-1016 (2012-08-23)
Dysfunction of PINK1, a mitochondrial Ser/Thr kinase, causes familial Parkinson's disease (PD). Recent studies have revealed that PINK1 is rapidly degraded in healthy mitochondria but accumulates on the membrane potential (ΔΨm)-deficient mitochondria, where it recruits another familial PD gene product
Grace G Y Lim et al.
PloS one, 8(9), e73235-e73235 (2013-09-12)
Disruption of the ubiquitin-proteasome system, which normally identifies and degrades unwanted intracellular proteins, is thought to underlie neurodegeneration. Supporting this, mutations of Parkin, a ubiquitin ligase, are associated with autosomal recessive parkinsonism. Remarkably, Parkin can protect neurons against a wide
Transition Metal Complexes of Ethopropazine: Synthesis and Characterization
Debbeti V, et al.
American Journal of Chemistry, 2(6), 294-298 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service