Skip to Content
Merck
All Photos(2)

Key Documents

E3635

Sigma-Aldrich

cis-11-Eicosenoic acid

≥99% (capillary GC)

Synonym(s):

cis-Gondoic acid, Gondoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)9CO2H
CAS Number:
Molecular Weight:
310.51
Beilstein:
1727313
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

plant oil (Jojoba)

Assay

≥99% (capillary GC)

form

liquid

refractive index

n20/D 1.4606 (lit.)

mp

23-24 °C (lit.)

density

0.883 g/mL at 25 °C (lit.)

functional group

carboxylic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCCC(O)=O

InChI

1S/C20H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h9-10H,2-8,11-19H2,1H3,(H,21,22)/b10-9-

InChI key

BITHHVVYSMSWAG-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Characteristics of Purified Horse Oil by Supercritical Fluid Extraction with Different Deodorants Agents.: This study investigates the characteristics of horse oil purified using supercritical fluid extraction and evaluates the efficacy of different deodorant agents. The research highlights the potential of cis-11-Eicosenoic acid in improving the quality and stability of horse oil, which has applications in food science and cosmetic industries (Anneke et al., 2024).

  • NEFA can serve as good biological markers for the diagnosis of depression in adolescents.: This research identifies non-esterified fatty acids (NEFA), including cis-11-Eicosenoic acid, as potential biomarkers for diagnosing depression in adolescents. The study provides insights into the biochemical pathways involved in mental health and the potential for developing new diagnostic tools (Liu et al., 2024).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Xu et al.
STAR protocols, 2(3), 100665-100665 (2021-07-27)
Mutations in microglia may cause brain disorders. Replacement of dysfunctional microglia by allogeneic wild-type microglia from bone marrow transplantation (Mr BMT) or peripheral blood can correct the gene deficiency at the brain-wide scale but cannot achieve precise replacement at specific
Guan-yuan Chen et al.
Clinica chimica acta; international journal of clinical chemistry, 438, 126-134 (2014-08-26)
As fatty acids play an important role in biological regulation, the profiling of fatty acid expression has been used to discover various disease markers and to understand disease mechanisms. This study developed an effective and accurate comparative fatty acid analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service