Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benzonase
®
Nuclease
Synonym(s):
Endonuclease from
Serratia marcescens
CAS Number:
9025-65-4
Products
Product Category
enzymes (3)
Brand
Millipore (3)
Biological Source
Physical Form
Purity
Recombinant Host
Quality Segment
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
E1014
Benzonase
®
Nuclease,
≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in
E. coli
, buffered aqueous glycerol solution
Expand
View Pricing
70664
Benzonase
®
Nuclease,
Purity > 99%, Effective viscosity reduction and removal of nucleic acids from protein solutions
Expand
View Pricing
70746
Benzonase
®
Nuclease,
Purity > 99%
Expand
View Pricing