Skip to Content
Merck
All Photos(1)

Key Documents

E0789

Sigma-Aldrich

Casein, Hammarsten bovine

Synonym(s):

Casein from bovine milk

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

bovine milk (Hammarsten)

Assay

>90% dry basis

form

powder

technique(s)

enzyme immunoassay: suitable

mp

280 °C (dec.) (lit.)

UniProt accession no.

storage temp.

room temp

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Casein is an amphiphilic protein.
Casein, a phosphoprotein, is the principal protein component of milk which is almost entirely expressed in the lactating mammary gland. In bovine milk they are classified according to their calcium sensitivity: αs1-, αs2-, and β-caseins precipitate in the presence of calcium; whereas κ-casein remains in solution.

Application

Casein, Hammarsten bovine has been used as a substrate for Aplysia californica calpain to study its activity.
The product is useful as a protease substrate.

Biochem/physiol Actions

Casein is useful in food industries and non-food applications. It has the property for emulsification, foam formation, and stabilization, water-binding, and gelation. Casein is also considered heat and acid-stable.
Partial gastrointestinal digestion of casein is a rich source of bioactive peptides, such as β-casomorphin. However, bovine casein is not homogeneous; variants A1 and B do lead to production of β-casomorphin 7 production, while A2 does not.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaret H Hastings et al.
PloS one, 12(10), e0186646-e0186646 (2017-10-21)
Calpains are a family of intracellular proteases defined by a conserved protease domain. In the marine mollusk Aplysia californica, calpains are important for the induction of long-term synaptic plasticity and memory, at least in part by cleaving protein kinase Cs
Krisztina Nagy et al.
The Journal of biological chemistry, 285(50), 38811-38817 (2010-10-06)
The possible mechanism of casein aggregation and micelle buildup was studied in a new approach by letting α-casein adsorb from low concentration (0.1 mg·ml(-1)) solutions onto the charged surfaces of polyelectrolyte films. It was found that α-casein could adsorb onto
Andreas F Kolb et al.
PloS one, 6(7), e21775-e21775 (2011-07-27)
The major physiological function of milk is the transport of amino acids, carbohydrates, lipids and minerals to mammalian offspring. Caseins, the major milk proteins, are secreted in the form of a micelle consisting of protein and calcium-phosphate.We have analysed the
Bovine beta-casein: Isolation, properties and functionality. A review
Atamer Z, et al.
International dairy journal, 66, 115-125 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service