Skip to Content
Merck
All Photos(1)

Key Documents

D9302

Sigma-Aldrich

Diacerein

≥95% (HPLC)

Synonym(s):

1,8-Diacetoxy-3-carboxyanthraquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12O8
CAS Number:
Molecular Weight:
368.29
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

powder

SMILES string

CC(=O)Oc1cccc2C(=O)c3cc(cc(OC(C)=O)c3C(=O)c12)C(O)=O

InChI

1S/C19H12O8/c1-8(20)26-13-5-3-4-11-15(13)18(23)16-12(17(11)22)6-10(19(24)25)7-14(16)27-9(2)21/h3-7H,1-2H3,(H,24,25)

InChI key

TYNLGDBUJLVSMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Diacerin is an inhibitor of Interleukin-1B (IL-1B) production.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Bartels et al.
Osteoarthritis and cartilage, 18(3), 289-296 (2009-10-28)
To estimate the efficacy and safety of diacerein as a pain-reducing agent in the treatment of osteoarthritis (OA), using meta-analysis of published randomized placebo-controlled trials (RCTs). Systematic searches of the bibliographic databases Medline, Embase, Cinahl, Chemical Abstracts, Cochrane and Web
Pascal Richette
Therapie, 66(5), 383-390 (2011-10-28)
According to international recommendations, acetaminophen should be used as a first-line therapy in patients with osteoarthritis, because of its safety and effectiveness. NSAIDs should be considered in patients unresponsive to acetaminophen, and should be prescribed at the lowest effective dose
Vinícius Maria Gadotti et al.
Pharmacology, biochemistry, and behavior, 102(4), 549-554 (2012-07-04)
The present study evaluated the antinociceptive effect of the pro-inflammatory cytokines inhibitor diacerein in mice and its possible mechanism of action. The antinociception produced by diacerein was tested at different sites of action, moreover selective antagonists or agonists were used
P Härle et al.
Der Internist, 49(12), 1458-1462 (2008-11-18)
There is an increasing incidence in osteoarthritis, particularly following the 5th life-decade. However, also young people may suffer from severe osteoarthritis, which is estimated to be the most common cause of disability in adults resulting in substantial economic burden. To
Umut Kalyoncu et al.
Joint, bone, spine : revue du rhumatisme, 76(4), 389-393 (2009-02-17)
The objective of the present study was to assess the frequency of self-reported psoriasis in a hip osteoarthritis (OA) cohort, and a secondary objective was to assess the course of hip OA with psoriasis. ECHODIAH was a 3-year, randomised double-blind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service