Skip to Content
Merck
All Photos(1)

Documents

D6196

Sigma-Aldrich

Dimebon dihydrochloride hydrate

≥98% (HPLC)

Synonym(s):

2,8-Dimethyl-5-[2-(6-methyl-3-pyridinyl)ethyl]-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole dihydrochloride hydrate, Dimebolin dihydrochloride hydrate, Dimeboline dihydrochloride hydrate, Dimebone dihydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N3 · 2HCl · xH2O
CAS Number:
Molecular Weight:
392.37 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.Cl.CN1CCc2c(C1)c3cc(C)ccc3n2CCc4ccc(C)nc4

InChI

1S/C21H25N3.2ClH.H2O/c1-15-4-7-20-18(12-15)19-14-23(3)10-9-21(19)24(20)11-8-17-6-5-16(2)22-13-17;;;/h4-7,12-13H,8-11,14H2,1-3H3;2*1H;1H2

InChI key

QQHQMWVYYVVAOV-UHFFFAOYSA-N

Biochem/physiol Actions

Mechanism of neuroprotection and cognition enhancement is not certain, but Dimebon blocks both cholinesterase and the NMDA receptor simultaneously and also appears to block the action of neurotoxic β-amyloid proteins and inhibit L-type calcium channels, and may exert a neuroprotective effect by blocking a novel target that involves mitochondrial pores.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Duarte et al.
Biomolecules, 12(2) (2022-02-26)
Despite the progressive research and recent advances in drug therapy to treat solid tumours, the number of cases and deaths in patients with cancer is still a major health problem. Drug repurposing coupled to drug combination strategies has been gaining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service