Skip to Content
Merck
All Photos(3)

Key Documents

D5042

Sigma-Aldrich

123 bp DNA Ladder

for DNA electrophoresis

Synonym(s):

DNA Ladder for DNA electrophoresis, DNA marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335
NACRES:
NA.25

Quality Level

form

liquid

usage

100 uses (0.1 mg size)
250 uses (250 μg size)

suitability

suitable for electrophoresis (DNA)

storage temp.

−20°C

Application

123 bp DNA Ladder has been used as a size marker in random amplified polymorphic DNA-polymerase chain reaction (PCR) analysis. It has also been used as a molecular marker in gel electrophoresis.

Components

Sigma′s 123 bp DNA Ladder is provided in a solution of 10 mM Tris-HCl (pH 7.5), 50 mM NaCl, and 0.1 mM EDTA.

Other Notes

Contains 35 fragments, 123 - 4182 bp
For optimal resolution, the recommended agarose gel concentration is 1.5%.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin-like growth factor I and II preserve myocardial structure in postinfarct swine
Kotlyar AA, et al.
British Heart Journal, 86(6), 693-700 (2001)
Straightforward procedure for laboratory production of DNA ladder
Lan VTT, et al.
Journal of Nucleic Acids, 2012 (2012)
Molecular characterization of lactic acid bacteria isolated from beef and stored using vacuum-packaging and advanced vacuum skin packaging systems
Inguane AS, et al.
CYTA-journal of Food (2011)
J L Hartley et al.
Gene, 13(4), 347-353 (1981-05-01)
A method for self-ligation of DNA segments, which is based on the rotational non-equivalence of ends produced by AvaI cleavage, has been developed. Using this method and an initiator molecule to encourage the formation of long polymers, we have obtained
A simple and practical method that prepares high molecular weight DNA ladders
Zhang JH, et al.
Molecular Medicine Reports, 6(5), 1211-1213 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service