Skip to Content
Merck
All Photos(2)

Key Documents

D3570

Sigma-Aldrich

Anti-DVL1 antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution, antigen mol wt ~85 kDa

Synonym(s):

Anti-Dishevelled-1, Anti-Dsh homolog 1, Anti-Segment polarity protein dishevelled homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~85 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cell lysate expressing human DVL1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DVL1(1855)

General description

The three DVL isoforms display high sequence homology and have conserved DIX, PDZ and DEP domains required for GSK3β inactivation. DVL1 is highly expressed in adult and fetal tissues such as skeletal muscle and pancreas but is also found in the brain and neural tube.

Application

Anti-DVL1 antibody produced in rabbit has been used in immunoblotting and immunohistochemistry (IHC).

Biochem/physiol Actions

Dishevelled (Dsh, DVL) proteins are part of a multigene family that mediate wnt signaling pathways that are essential for the regulation of cellular proliferation, differentiation, motility, and morphogenesis. DVL1 regulates the activity of JNK (c-Jun N-terminal kinase) and GSK3β in the wnt signaling pathway. Upon activation of the wnt signaling pathway, DVL1 inactivates GSK3β through complex formation with APC (Antigen-presenting cell), β-catenin and axin proteins, releasing β-catenin from degradation. In neurons, DVL1 is localized to axonal microtubules and stabilizes microtubules by inhibiting the GSK3β mediated phosphorylation of MAP-1B.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Chen et al.
International journal of clinical and experimental pathology, 6(9), 1791-1798 (2013-09-17)
Hirschsprung's disease (HSCR) is a congenital disorder of the enteric nervous system and is characterized by an absence of enteric ganglion cells in terminal regions of the gut during development. Dishevelled (DVL) protein is a cytoplasmic protein which plays pivotal
DVL1 and DVL3 differentially localize to CYP19A1 promoters and regulate aromatase mRNA in breast cancer cells
Castro PI, et al.
Oncotarget, 9(86), 35639-35639 (2018)
ALFY-Controlled DVL3 Autophagy Regulates Wnt Signaling, Determining Human Brain Size
Kadir R, et al.
PLoS Genetics, 12(3), e1005919-e1005919 (2016)
Monica Sharma et al.
Genes & cancer, 12, 77-95 (2021-10-19)
Dishevelled-1 (DVL-1) mediates Wnt signals critical for development and cellular homeostasis. DVL-1 is also linked with tumorigenesis, however its association with specific breast cancer (BC) subtypes and how it contributes to tumorigenicity remains poorly studied. Herein, using bioinformatics and genomics
Lorenza Ciani et al.
The Journal of cell biology, 164(2), 243-253 (2004-01-22)
Dishevelled (DVL) is associated with axonal microtubules and regulates microtubule stability through the inhibition of the serine/threonine kinase, glycogen synthase kinase 3beta (GSK-3beta). In the canonical WNT pathway, the negative regulator Axin forms a complex with beta-catenin and GSK-3beta, resulting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service