Skip to Content
Merck
All Photos(2)

Key Documents

D3190

Sigma-Aldrich

Dithiobiuret

97%, solid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

97%

form

solid

SMILES string

NC(=S)NC(N)=S

InChI

1S/C2H5N3S2/c3-1(6)5-2(4)7/h(H5,3,4,5,6,7)

InChI key

JIRRNZWTWJGJCT-UHFFFAOYSA-N

General description

Paralytic agent.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W D Atchison
The Journal of pharmacology and experimental therapeutics, 249(3), 735-743 (1989-06-01)
Daily treatment of rats with 2,4-dithiobiuret (DTB, 1 mg/kg/day i.p.) produces a flaccid neuromuscular weakness first observed in the hindlimbs after 5 to 6 days of treatment. This condition is characterized by diminished contractile strength following single shock and tetanic
W R Porter et al.
Neurotoxicology, 4(4), 57-68 (1983-01-01)
[14C] Dithiobiuret (DTB)-derived radioactivity is eliminated by adult male rats with an approximate plasma half-life of 8-10 hr. About 65-75% of an i.p. dose appears in the urine within 24 hr after treatment and about 2-4% appears in the feces
L M Ireland et al.
The Journal of pharmacology and experimental therapeutics, 275(3), 1453-1462 (1995-12-01)
Chronic administration of 2,4-dithiobiuret (DTB), causes delayed-onset neuromuscular weakness in rats. This effect results from inhibition of quantal release of acetylcholine (ACh) from motor nerve terminals. The effects of noncholinergic neurotransmission are unknown. The purpose of the present study was
H B Jones
Acta neuropathologica, 78(1), 72-85 (1989-01-01)
2,4-Dithiobiuret was given i.p. to rats for 4 days at a daily dosage of 1 mg/kg and the development of the lesion associated with neuromuscular dysfunction studied in hindlimb lumbrical muscles. The first morphological indication of neurointoxication was the appearance
W D Atchison
Toxicology and applied pharmacology, 106(2), 234-244 (1990-11-01)
Daily treatment of rats with 2,4-dithiobiuret (DTB, 1 mg/kg/day, ip) causes a flaccid neuromuscular weakness first observable in the hindlimbs after 5-6 days of treatment. With continued exposure, neuromuscular weakness appears to encompass the other muscles of the body; death

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service