Skip to Content
Merck
All Photos(2)

Documents

D1156

Sigma-Aldrich

N,N-Dimethylglycine

≥99%, suitable for ligand binding assays

Synonym(s):

(Dimethylamino)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2COOH
CAS Number:
Molecular Weight:
103.12
Beilstein:
1700261
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

N,N-Dimethylglycine, ≥99%

Quality Level

Assay

≥99%

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

mp

178-182 °C (lit.)

application(s)

peptide synthesis

SMILES string

CN(C)CC(O)=O

InChI

1S/C4H9NO2/c1-5(2)3-4(6)7/h3H2,1-2H3,(H,6,7)

InChI key

FFDGPVCHZBVARC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. N,N-Dimethylglycine is used in the development of glycine-based ionic liquids and emulsifiers. N,N-Dimethylglycine is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baljit K Ubhi et al.
The European respiratory journal, 40(2), 345-355 (2011-12-21)
There is a paucity of biomarkers for chronic obstructive pulmonary disease (COPD). Metabolomics were applied to a defined COPD patient cohort from the ECLIPSE study (Evaluation of COPD Longitudinally to Identify Predictive Surrogate End-points). Results were correlated with accepted biomarkers
S H Moolenaar et al.
Clinical chemistry, 45(4), 459-464 (1999-04-02)
A38-year-old man presented with a history of fish odor (since age 5) and unusual muscle fatigue with increased serum creatine kinase. Our aim was to identify the metabolic error in this new condition. We used 1H NMR spectroscopy to study
Ali A Moazzami et al.
The Journal of nutrition, 141(12), 2126-2132 (2011-10-21)
Prostate cancer (PC) is the most common cancer in the Western world and the second most important cancer causing male deaths, after lung cancer, in the United States and Britain. Lifestyle and dietary changes are recommended for men diagnosed with
D Vanhauteghem et al.
Amino acids, 43(1), 245-253 (2011-09-14)
The current study first investigates the emulsifying potential of glycine and its N-methylated derivatives N-methylglycine (sarcosine), N,N-dimethylglycine (DMG) and N,N,N-trimethylglycine (betaine) under varying pH conditions. Subsequently, the effect of these test compounds on the membrane integrity of enterotoxigenic Escherichia coli
Isabelle D Kalmar et al.
The British journal of nutrition, 107(11), 1635-1644 (2011-09-21)
N,N-Dimethylglycine (DMG) is a tertiary amino acid that naturally occurs as an intermediate metabolite in choline-to-glycine metabolism. The objective of the present trial was to evaluate tolerance, safety and bioaccumulation of dietary DMG in broilers when supplemented at 1 g

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service