Skip to Content
Merck
All Photos(3)

Key Documents

S7672

Sigma-Aldrich

Sarcosine

≥98%, suitable for HPLC, BioXtra

Synonym(s):

N-Methylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCH2COOH
CAS Number:
Molecular Weight:
89.09
Beilstein:
1699442
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

product name

Sarcosine, BioXtra

product line

BioXtra

Assay

≥98%

form

powder

technique(s)

HPLC: suitable

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

color

white

mp

208-212 °C (dec.) (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.5%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.1%
Na: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

application(s)

cell analysis

SMILES string

CNCC(O)=O

InChI

1S/C3H7NO2/c1-4-2-3(5)6/h4H,2H2,1H3,(H,5,6)

InChI key

FSYKKLYZXJSNPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Glyphosate resistance and biodegradation by Burkholderia cenocepacia CEIB S5-2.: This study explores the role of sarcosine in glyphosate resistance and biodegradation by Burkholderia cenocepacia CEIB S5-2, highlighting its potential in environmental biotechnology (Díaz-Soto et al., 2024).

  • CYP116B5-SOX: An artificial peroxygenase for drug metabolites production and bioremediation.: Sarcosine is utilized in developing a novel artificial peroxygenase, enhancing bioremediation and drug metabolite production applications (Giuriato et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiyi Wu et al.
The Journal of physiology, 598(16), 3417-3438 (2020-05-24)
Loss-of-function mutations in proteins found at glycinergic synapses, most commonly in the α1 subunit of the glycine receptor (GlyR), cause the startle disease/hyperekplexia channelopathy in man. It was recently proposed that the receptors responsible are presynaptic homomeric GlyRs, rather than
Florian Jentzmik et al.
The Journal of urology, 185(2), 706-711 (2010-12-21)
Sarcosine in prostate cancer tissue samples was recently reported to be increased during prostate cancer progression to metastasis and suggested to be a key metabolite of cancer cell invasion and aggressiveness. We reevaluated sarcosine in prostate cancer tissue samples as
D S Lim et al.
Journal of the American College of Cardiology, 38(4), 1175-1180 (2001-10-05)
The goal of this study was to identify genes upregulated in the heart in human patients with hypertrophic cardiomyopathy (HCM). Hypertrophic cardiomyopathy is a genetic disease caused by mutations in contractile sarcomeric proteins. The molecular basis of diverse clinical and
Eri Hara et al.
Biochimica et biophysica acta, 1830(8), 4046-4052 (2013-04-03)
Nanoparticle of Lactosome, which is composed of poly(l-lactic acid)-base depsipeptide with diameter of 35nm, accumulates in solid tumors by the enhanced permeability and retention (EPR) effect. However, a pharmacokinetic alteration of Lactosome was observed when Lactosome was repeatedly administered. This
J R Bales et al.
Clinical chemistry, 30(10), 1631-1636 (1984-10-01)
Acetaminophen and its glucuronide, sulfate, N-acetyl-L-cysteinyl, and L-cysteinyl metabolites can be rapidly detected by 1H NMR spectroscopy of intact, untreated human urine. Study of the time course of excretion of these metabolites in five clinically normal men after ingestion of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service