Skip to Content
Merck
All Photos(2)

Key Documents

D0676

Sigma-Aldrich

Dobutamine hydrochloride

≥98% (TLC), powder, α1, β1 and β2-adrenoceptor agonist

Synonym(s):

(±)-3,4-Dihydroxy-N-[3-(4-hydroxyphenyl)-1-methylpropyl]-β-phenethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO3 · HCl
CAS Number:
Molecular Weight:
337.84
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Product Name

Dobutamine hydrochloride, ≥98%

Quality Level

Assay

≥98%

form

powder

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

Cl[H].CC(CCc1ccc(O)cc1)NCCc2ccc(O)c(O)c2

InChI

1S/C18H23NO3.ClH/c1-13(2-3-14-4-7-16(20)8-5-14)19-11-10-15-6-9-17(21)18(22)12-15;/h4-9,12-13,19-22H,2-3,10-11H2,1H3;1H

InChI key

BQKADKWNRWCIJL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Dobutamine hydrochloride has been used:
  • to treat mouse embryonic fibroblasts for cell death analysis
  • to inhibit TAZ (transcriptional coactivator with PDZ-binding motif)/YAP (Yes-associated protein) of Hippo pathway and study its effect on tumor cell proliferation
  • as β1-AR partial agonist to determine its effects on budding of endophilin-mRFP-positive structures by live-cell imaging

Biochem/physiol Actions

Dobutamine is a synthetic catecholamine that stimulate the β1 -adrenoceptor receptor. It exhibits inotropic outcome on the myocardium. Dobutamine elevates contractility and is known to be useful in the condition of low blood pressure.

Features and Benefits

This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of p53 in cAMP/PKA pathway mediated apoptosis
Rahimi A, et al.
Apoptosis, 18(12), 1492-1499 (2013)
Workbook in Practical Neonatology, 239-239 (2014)
Programming of Schwann cells by Lats1/2-TAZ/YAP signaling drives malignant peripheral nerve sheath tumorigenesis
Wu LMN, et al.
Cancer Cell, 33(2), 292-308 (2018)
Dobutamine (2018)
Anumeha Tandon et al.
JACC. Cardiovascular imaging, 6(2), 184-195 (2013-03-16)
Although most patients with severe aortic stenosis (AS) have high peak velocity and mean transvalvular gradient, there is a subset of patients with low-flow, low-gradient severe AS (LGSAS). Assessment and management of such patients can be difficult and dobutamine echocardiography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service