Skip to Content
Merck

Skip To

CS2000

SARS-CoV-2 (COVID-19) Inhibitors Screening Kit 

Based on SPIKE - ACE2 Binding Assay, Colorimetric, sufficient for 200 tests

Synonym(s):

ACE2, COVID-19, S protein, SARS-CoV-2, Spike protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.84
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

shipped in

ambient

storage temp.

2-8°C

General description

designed to simplify the identification and characterization of SARS-CoV-2 inhibitors.

Application

The SARS-CoV-2 Inhibitors Screening Kit is a convenient tool, designed to simplify the identification and characterization of SARS-CoV-2 inhibitors.The crutial interaction between the SARS-CoV-2 and human cells accrues through the protein-protein interaction of human receptor ACE2 and viral protein Spike – S protein. The receptor binding domain (RBD) of S protein is the domain that specifically binds to ACE2. Our SARS-CoV-2 Inhibitors Screening Kit includes both human ACE protein and viral RBD of S protein. The kit is based on a colorimetric ELISA, where biotinylated ACE2 (ACE2-biotin) and Extravidin®-peroxidase are used to detect the ACE2:RBD interaction. One molecule of Extravidin® binds four molecules of biotin by a non-covalent interaction that is essentially irreversible. Binding visualization is achieved by using o-Phenylenediamine dihydrochloride (OPD), a peroxidase substrate with absorbance at 450 nm.

Inhibitors and neutralizing antibodies against RBD that interfere with the ACE2:RBD interaction will result in reduced colorimetric signal.

Biochem/physiol Actions

SARS-CoV-2 - Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) belongs to Coronaviridae (CoVs) - a large family of viruses that usually cause mild to moderate upper-respiratory tract diseases.1 CoVs are single-stranded RNA viruses. They have a high mutation rate and are therefore highly diverse.1 Their hosts include human and non-human mammals, as well as birds.1

SARS-CoV-2 infects cells which express angiotensin converting enzyme 2 (ACE2).2 ACE2 binds to the viral envelope-associated spike (S) protein as a cellular receptor, following proteolytic cleavage of both S and ACE2 by serine proteases.3 S protein is believed to be the target of neutralizing antibodies because it is the main trans-membrane glycoprotein responsible for receptor-binding and virion entry.4

The S protein is a trimeric protein. During the binding to ACE2, S protein undergoes a substantial structural rearrangement to facilitate the fusion of the viral membrane with the host cell membrane.5 The receptor binding domain (RBD) of S protein is the domain that specifically binds to ACE2. RBD is the major target for neutralizing antibodies for coronaviruses6, and anti-RBD antibodies likely correlate with virus neutralization.7

Features and Benefits

Our SARS-CoV-2 Inhibitors Screening Kit will support your research, enabling to screen and discover inhibitors as potential drugs for COVID-19
Supportive calculator (Click here to download a calculator excel file)

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAE2002SAE1000SAE2001
Quality Level

200

Quality Level

300

Quality Level

300

Quality Level

300

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

ambient

shipped in

-

shipped in

ambient

shipped in

-


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters